Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66272030-66272303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB1B
Entrez Gene ID
81876
Gene Name
RAB1B, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.300816
Gene Chromosome Location
Chr.11: 66268526-66277492 on Build GRCh38
Transcripts
NM_030981.2 XM_017018378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3810127 c.128T>G p.V43G Chr.11: 66272197-66272197 on Build GRCh38
5373310 c.92A>T p.D31V Chr.11: 66272161-66272161 on Build GRCh38
6761296 c.124G>A p.G42R Chr.11: 66272193-66272193 on Build GRCh38
7190811 c.106A>C p.S36R Chr.11: 66272175-66272175 on Build GRCh38
5500024 c.172A>C p.K58Q Chr.11: 66272241-66272241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776669430 Chr.11: 66272222-66272222 on Build GRCh38
rs748997758 Chr.11: 66272165-66272165 on Build GRCh38
rs751372119 Chr.11: 66272063-66272063 on Build GRCh38
rs986695235 Chr.11: 66272268-66272268 on Build GRCh38
rs774008831 Chr.11: 66272177-66272177 on Build GRCh38
rs765236066 Chr.11: 66272252-66272252 on Build GRCh38
rs755825172 Chr.11: 66272270-66272270 on Build GRCh38
rs575125265 Chr.11: 66272066-66272066 on Build GRCh38
rs561758693 Chr.11: 66272262-66272262 on Build GRCh38
rs745720151 Chr.11: 66272136-66272136 on Build GRCh38
rs780959402 Chr.11: 66272120-66272120 on Build GRCh38
rs980717207 Chr.11: 66272088-66272088 on Build GRCh38
rs925153944 Chr.11: 66272264-66272264 on Build GRCh38
rs766798368 Chr.11: 66272216-66272216 on Build GRCh38
rs1024987177 Chr.11: 66272052-66272052 on Build GRCh38
rs200047760 Chr.11: 66272143-66272143 on Build GRCh38
rs761693726 Chr.11: 66272138-66272138 on Build GRCh38
rs773692622 Chr.11: 66272201-66272201 on Build GRCh38
rs372615716 Chr.11: 66272261-66272261 on Build GRCh38
rs369083319 Chr.11: 66272279-66272279 on Build GRCh38
rs921245852 Chr.11: 66272070-66272070 on Build GRCh38
rs750197542 Chr.11: 66272263-66272263 on Build GRCh38
rs772767395 Chr.11: 66272198-66272198 on Build GRCh38
rs767999265 Chr.11: 66272171-66272171 on Build GRCh38
rs759820359 Chr.11: 66272226-66272226 on Build GRCh38
rs779608788 Chr.11: 66272144-66272144 on Build GRCh38
rs754729470 Chr.11: 66272281-66272281 on Build GRCh38
rs540836609 Chr.11: 66272111-66272111 on Build GRCh38
rs748663907 Chr.11: 66272192-66272192 on Build GRCh38
rs767613623 Chr.11: 66272259-66272259 on Build GRCh38
rs764163346 Chr.11: 66272062-66272062 on Build GRCh38
rs779919661 Chr.11: 66272272-66272272 on Build GRCh38
rs369688518 Chr.11: 66272204-66272204 on Build GRCh38

View Full Product Details