Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78972238-78972511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGALS3BP
Entrez Gene ID
3959
Gene Name
galectin 3 binding protein
Gene Aliases
90K, BTBD17B, CyCAP, M2BP, MAC-2-BP, TANGO10B, gp90
UniGene
Hs.514535
Gene Chromosome Location
Chr.17: 78971253-78979979 on Build GRCh38
Transcripts
NM_005567.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318158 c.1012G>A p.E338K Chr.17: 78972322-78972322 on Build GRCh38
5903150 c.854G>A p.W285* Chr.17: 78972480-78972480 on Build GRCh38
1256414 c.856A>G p.N286D Chr.17: 78972478-78972478 on Build GRCh38
4405643 c.1010A>C p.E337A Chr.17: 78972324-78972324 on Build GRCh38
3717719 c.965T>A p.L322Q Chr.17: 78972369-78972369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978194431 Chr.17: 78972395-78972395 on Build GRCh38
rs528455834 Chr.17: 78972462-78972462 on Build GRCh38
rs764292028 Chr.17: 78972392-78972392 on Build GRCh38
rs138118783 Chr.17: 78972473-78972473 on Build GRCh38
rs953178025 Chr.17: 78972354-78972354 on Build GRCh38
rs371295281 Chr.17: 78972442-78972442 on Build GRCh38
rs773459748 Chr.17: 78972295-78972295 on Build GRCh38
rs529673767 Chr.17: 78972422-78972422 on Build GRCh38
rs752735923 Chr.17: 78972317-78972317 on Build GRCh38
rs772172848 Chr.17: 78972361-78972361 on Build GRCh38
rs750176552 Chr.17: 78972419-78972419 on Build GRCh38
rs371251313 Chr.17: 78972328-78972328 on Build GRCh38
rs762096995 Chr.17: 78972484-78972484 on Build GRCh38
rs776881671 Chr.17: 78972397-78972397 on Build GRCh38
rs764320304 Chr.17: 78972320-78972320 on Build GRCh38
rs776532542 Chr.17: 78972483-78972483 on Build GRCh38
rs780891202 Chr.17: 78972352-78972352 on Build GRCh38
rs948285330 Chr.17: 78972451-78972451 on Build GRCh38
rs3803782 Chr.17: 78972455-78972455 on Build GRCh38
rs760894840 Chr.17: 78972318-78972318 on Build GRCh38
rs572007354 Chr.17: 78972362-78972362 on Build GRCh38
rs368284637 Chr.17: 78972327-78972327 on Build GRCh38
rs771404500 Chr.17: 78972306-78972306 on Build GRCh38
rs763361258 Chr.17: 78972304-78972304 on Build GRCh38
rs188404201 Chr.17: 78972312-78972312 on Build GRCh38
rs916807930 Chr.17: 78972448-78972448 on Build GRCh38
rs201912427 Chr.17: 78972282-78972282 on Build GRCh38
rs374500621 Chr.17: 78972338-78972338 on Build GRCh38
rs202012874 Chr.17: 78972336-78972336 on Build GRCh38
rs886217844 Chr.17: 78972469-78972469 on Build GRCh38
rs765318272 Chr.17: 78972490-78972490 on Build GRCh38
rs773199042 Chr.17: 78972491-78972491 on Build GRCh38
rs541114993 Chr.17: 78972383-78972383 on Build GRCh38
rs370838910 Chr.17: 78972472-78972472 on Build GRCh38
rs549735080 Chr.17: 78972424-78972424 on Build GRCh38
rs779629155 Chr.17: 78972374-78972374 on Build GRCh38
rs778757224 Chr.17: 78972334-78972334 on Build GRCh38
rs182994096 Chr.17: 78972307-78972307 on Build GRCh38
rs768355767 Chr.17: 78972294-78972294 on Build GRCh38
rs149291165 Chr.17: 78972400-78972400 on Build GRCh38
rs368741094 Chr.17: 78972428-78972428 on Build GRCh38
rs199912199 Chr.17: 78972308-78972308 on Build GRCh38
rs747815135 Chr.17: 78972355-78972355 on Build GRCh38
rs754474890 Chr.17: 78972434-78972434 on Build GRCh38
rs768248593 Chr.17: 78972344-78972344 on Build GRCh38
rs772503008 Chr.17: 78972382-78972382 on Build GRCh38
rs536183507 Chr.17: 78972326-78972326 on Build GRCh38
rs141960783 Chr.17: 78972480-78972480 on Build GRCh38
rs564643221 Chr.17: 78972337-78972337 on Build GRCh38
rs560765759 Chr.17: 78972401-78972401 on Build GRCh38
rs61734154 Chr.17: 78972335-78972335 on Build GRCh38
rs771677674 Chr.17: 78972258-78972258 on Build GRCh38
rs534200430 Chr.17: 78972284-78972284 on Build GRCh38
rs779256192 Chr.17: 78972461-78972461 on Build GRCh38
rs746689807 Chr.17: 78972264-78972264 on Build GRCh38
rs760731529 Chr.17: 78972388-78972388 on Build GRCh38
rs144649043 Chr.17: 78972454-78972454 on Build GRCh38

View Full Product Details