Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32713320-32713569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1L
Entrez Gene ID
10059
Gene Name
dynamin 1 like
Gene Aliases
DLP1, DRP1, DVLP, DYMPLE, EMPF, HDYNIV
UniGene
Hs.556296
Gene Chromosome Location
Chr.12: 32679200-32745650 on Build GRCh38
Transcripts
NM_001278463.1 NM_001278464.1 NM_001278465.1 NM_001278466.1 NM_005690.4 NM_012062.4 NM_012063.3 XM_005253282.4 XM_011520543.2 XM_011520544.2 XM_017018663.1 XM_017018664.1 XM_017018665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3460222 c.629T>C p.L210P Chr.12: 32713342-32713342 on Build GRCh38
3460221 c.590T>C p.L197P Chr.12: 32713342-32713342 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs367668026 Chr.12: 32713395-32713395 on Build GRCh38
rs945095735 Chr.12: 32713441-32713441 on Build GRCh38
rs7971495 Chr.12: 32713465-32713465 on Build GRCh38
rs760798591 Chr.12: 32713457-32713457 on Build GRCh38
rs772027996 Chr.12: 32713423-32713423 on Build GRCh38
rs375929274 Chr.12: 32713367-32713367 on Build GRCh38
rs762111681 Chr.12: 32713476-32713476 on Build GRCh38
rs749615882 Chr.12: 32713365-32713365 on Build GRCh38
rs769177492 Chr.12: 32713370-32713370 on Build GRCh38
rs775615240 Chr.12: 32713378-32713378 on Build GRCh38
rs751135269 Chr.12: 32713514-32713514 on Build GRCh38
rs765658532 Chr.12: 32713489-32713489 on Build GRCh38
rs140398302 Chr.12: 32713502-32713502 on Build GRCh38
rs913270088 Chr.12: 32713463-32713463 on Build GRCh38
rs748289184 Chr.12: 32713385-32713385 on Build GRCh38
rs1042158534 Chr.12: 32713449-32713452 on Build GRCh38
rs7958074 Chr.12: 32713469-32713469 on Build GRCh38
rs957480374 Chr.12: 32713389-32713389 on Build GRCh38
rs780252977 Chr.12: 32713364-32713364 on Build GRCh38

View Full Product Details