Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139561648-139561921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F9
Entrez Gene ID
2158
Gene Name
coagulation factor IX
Gene Aliases
F9 p22, FIX, HEMB, P19, PTC, THPH8
UniGene
Hs.522798
Gene Chromosome Location
Chr.X: 139530720-139563459 on Build GRCh38
Transcripts
NM_000133.3 NM_001313913.1 XM_005262397.4

SNP Detail

SNP ID Chromosome Location
rs137852261 Chr.X: 139561835-139561835 on Build GRCh38
rs954954812 Chr.X: 139561711-139561711 on Build GRCh38
rs769827536 Chr.X: 139561723-139561723 on Build GRCh38
rs145026483 Chr.X: 139561714-139561714 on Build GRCh38
rs387906482 Chr.X: 139561716-139561716 on Build GRCh38
rs137852253 Chr.X: 139561694-139561694 on Build GRCh38
rs137852275 Chr.X: 139561755-139561755 on Build GRCh38
rs747941750 Chr.X: 139561861-139561861 on Build GRCh38
rs137852273 Chr.X: 139561872-139561872 on Build GRCh38
rs137852271 Chr.X: 139561805-139561805 on Build GRCh38
rs147567879 Chr.X: 139561733-139561733 on Build GRCh38
rs137852256 Chr.X: 139561749-139561749 on Build GRCh38
rs137852282 Chr.X: 139561854-139561854 on Build GRCh38
rs900302000 Chr.X: 139561894-139561894 on Build GRCh38
rs137852259 Chr.X: 139561821-139561821 on Build GRCh38
rs34698779 Chr.X: 139561795-139561795 on Build GRCh38
rs137852260 Chr.X: 139561829-139561829 on Build GRCh38
rs137852252 Chr.X: 139561683-139561683 on Build GRCh38
rs775836977 Chr.X: 139561724-139561724 on Build GRCh38
rs758194285 Chr.X: 139561875-139561875 on Build GRCh38
rs377350610 Chr.X: 139561839-139561839 on Build GRCh38
rs112057482 Chr.X: 139561780-139561780 on Build GRCh38
rs761784613 Chr.X: 139561828-139561828 on Build GRCh38
rs137852255 Chr.X: 139561743-139561743 on Build GRCh38
rs764515994 Chr.X: 139561739-139561739 on Build GRCh38
rs387906479 Chr.X: 139561773-139561773 on Build GRCh38
rs137852254 Chr.X: 139561710-139561710 on Build GRCh38
rs137852281 Chr.X: 139561874-139561874 on Build GRCh38
rs886038243 Chr.X: 139561717-139561717 on Build GRCh38
rs751992311 Chr.X: 139561759-139561759 on Build GRCh38
rs137852262 Chr.X: 139561865-139561865 on Build GRCh38
rs137852258 Chr.X: 139561820-139561820 on Build GRCh38
rs750955240 Chr.X: 139561792-139561792 on Build GRCh38
rs925187679 Chr.X: 139561726-139561726 on Build GRCh38
rs137852257 Chr.X: 139561754-139561754 on Build GRCh38
rs776509489 Chr.X: 139561676-139561676 on Build GRCh38
rs778623966 Chr.X: 139561850-139561850 on Build GRCh38
rs113427375 Chr.X: 139561777-139561777 on Build GRCh38
rs777757344 Chr.X: 139561876-139561876 on Build GRCh38
rs137852283 Chr.X: 139561836-139561836 on Build GRCh38

View Full Product Details