Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93524072-93524303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNBP1L
Entrez Gene ID
54874
Gene Name
formin binding protein 1 like
Gene Aliases
C1orf39, TOCA1
UniGene
Hs.134060
Gene Chromosome Location
Chr.1: 93448067-93569671 on Build GRCh38
Transcripts
NM_001024948.2 NM_001164473.2 NM_017737.4 XM_011541625.2 XM_017001533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418515 c.346C>A p.L116M Chr.1: 93524264-93524264 on Build GRCh38
4446001 c.343-3C>T p.? Chr.1: 93524258-93524258 on Build GRCh38
4446002 c.343-3C>T p.? Chr.1: 93524258-93524258 on Build GRCh38
6418514 c.346C>A p.L116M Chr.1: 93524264-93524264 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs973762227 Chr.1: 93524276-93524276 on Build GRCh38
rs773936682 Chr.1: 93524231-93524231 on Build GRCh38
rs758950022 Chr.1: 93524232-93524232 on Build GRCh38
rs997814341 Chr.1: 93524231-93524231 on Build GRCh38
rs1007050292 Chr.1: 93524151-93524151 on Build GRCh38
rs762998074 Chr.1: 93524193-93524193 on Build GRCh38
rs771801431 Chr.1: 93524231-93524232 on Build GRCh38
rs766343399 Chr.1: 93524243-93524243 on Build GRCh38
rs1031154618 Chr.1: 93524165-93524165 on Build GRCh38
rs1025335175 Chr.1: 93524232-93524232 on Build GRCh38
rs969420380 Chr.1: 93524261-93524261 on Build GRCh38
rs1039491698 Chr.1: 93524207-93524207 on Build GRCh38
rs527796664 Chr.1: 93524208-93524208 on Build GRCh38
rs752665163 Chr.1: 93524214-93524226 on Build GRCh38
rs936270738 Chr.1: 93524123-93524123 on Build GRCh38
rs150379851 Chr.1: 93524262-93524262 on Build GRCh38
rs896758117 Chr.1: 93524179-93524179 on Build GRCh38
rs549102008 Chr.1: 93524244-93524244 on Build GRCh38
rs889314746 Chr.1: 93524135-93524135 on Build GRCh38
rs1054540932 Chr.1: 93524128-93524128 on Build GRCh38

View Full Product Details