Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206842438-206842711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL19
Entrez Gene ID
29949
Gene Name
interleukin 19
Gene Aliases
IL-10C, MDA1, NG.1, ZMDA1
UniGene
Hs.661017
Gene Chromosome Location
Chr.1: 206798870-206842981 on Build GRCh38
Transcripts
NM_013371.3 NM_153758.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5438388 c.443A>C p.E148A Chr.1: 206842531-206842531 on Build GRCh38
1689744 c.514G>A p.E172K Chr.1: 206842602-206842602 on Build GRCh38
4837928 c.527C>T p.S176L Chr.1: 206842615-206842615 on Build GRCh38
1689743 c.628G>A p.E210K Chr.1: 206842602-206842602 on Build GRCh38
77887 c.481G>A p.D161N Chr.1: 206842569-206842569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757767065 Chr.1: 206842660-206842660 on Build GRCh38
rs772096252 Chr.1: 206842491-206842491 on Build GRCh38
rs1026485705 Chr.1: 206842466-206842466 on Build GRCh38
rs754068069 Chr.1: 206842481-206842481 on Build GRCh38
rs144719259 Chr.1: 206842569-206842569 on Build GRCh38
rs370571915 Chr.1: 206842614-206842614 on Build GRCh38
rs182270228 Chr.1: 206842637-206842637 on Build GRCh38
rs774399442 Chr.1: 206842599-206842599 on Build GRCh38
rs964764168 Chr.1: 206842526-206842526 on Build GRCh38
rs763055272 Chr.1: 206842642-206842642 on Build GRCh38
rs372603560 Chr.1: 206842632-206842632 on Build GRCh38
rs986608382 Chr.1: 206842480-206842480 on Build GRCh38
rs761691920 Chr.1: 206842461-206842461 on Build GRCh38
rs1054097136 Chr.1: 206842565-206842565 on Build GRCh38
rs905592787 Chr.1: 206842650-206842650 on Build GRCh38
rs371050229 Chr.1: 206842670-206842670 on Build GRCh38
rs773126530 Chr.1: 206842596-206842597 on Build GRCh38
rs752042997 Chr.1: 206842659-206842659 on Build GRCh38
rs540963634 Chr.1: 206842568-206842568 on Build GRCh38
rs186540222 Chr.1: 206842669-206842669 on Build GRCh38
rs372751115 Chr.1: 206842496-206842496 on Build GRCh38
rs779555269 Chr.1: 206842574-206842574 on Build GRCh38
rs375976152 Chr.1: 206842539-206842539 on Build GRCh38
rs913076620 Chr.1: 206842598-206842598 on Build GRCh38
rs745525215 Chr.1: 206842571-206842571 on Build GRCh38
rs761891620 Chr.1: 206842608-206842608 on Build GRCh38
rs768672561 Chr.1: 206842587-206842587 on Build GRCh38
rs764202554 Chr.1: 206842646-206842646 on Build GRCh38
rs748827538 Chr.1: 206842577-206842577 on Build GRCh38
rs758846775 Chr.1: 206842532-206842532 on Build GRCh38
rs560760235 Chr.1: 206842572-206842572 on Build GRCh38
rs765355428 Chr.1: 206842511-206842511 on Build GRCh38
rs2243191 Chr.1: 206842612-206842612 on Build GRCh38
rs781549106 Chr.1: 206842662-206842662 on Build GRCh38
rs778276774 Chr.1: 206842538-206842538 on Build GRCh38
rs752724737 Chr.1: 206842515-206842515 on Build GRCh38

View Full Product Details