Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23893120-23893379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKIRAS1
Entrez Gene ID
28512
Gene Name
NFKB inhibitor interacting Ras like 1
Gene Aliases
KBRAS1, kappaB-Ras1
UniGene
Hs.173202
Gene Chromosome Location
Chr.3: 23889536-23917046 on Build GRCh38
Transcripts
NM_020345.3 XM_005265076.3 XM_005265077.4 XM_005265078.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1043322 c.461G>A p.R154Q Chr.3: 23893213-23893213 on Build GRCh38
6726542 c.367G>A p.D123N Chr.3: 23893307-23893307 on Build GRCh38
117601 c.430G>A p.V144I Chr.3: 23893244-23893244 on Build GRCh38
4406219 c.371T>C p.L124P Chr.3: 23893303-23893303 on Build GRCh38
730412 c.516G>C p.Q172H Chr.3: 23893158-23893158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144795972 Chr.3: 23893157-23893157 on Build GRCh38
rs766818239 Chr.3: 23893305-23893305 on Build GRCh38
rs765604747 Chr.3: 23893173-23893174 on Build GRCh38
rs202127637 Chr.3: 23893148-23893148 on Build GRCh38
rs775297949 Chr.3: 23893268-23893268 on Build GRCh38
rs781307646 Chr.3: 23893213-23893213 on Build GRCh38
rs764845003 Chr.3: 23893288-23893288 on Build GRCh38
rs747509825 Chr.3: 23893191-23893191 on Build GRCh38
rs749476852 Chr.3: 23893344-23893344 on Build GRCh38
rs763537059 Chr.3: 23893310-23893311 on Build GRCh38
rs762725417 Chr.3: 23893293-23893293 on Build GRCh38
rs529275413 Chr.3: 23893277-23893277 on Build GRCh38
rs755477564 Chr.3: 23893206-23893206 on Build GRCh38
rs374026293 Chr.3: 23893214-23893214 on Build GRCh38
rs148321857 Chr.3: 23893221-23893221 on Build GRCh38
rs1031279337 Chr.3: 23893307-23893307 on Build GRCh38
rs919038413 Chr.3: 23893266-23893266 on Build GRCh38
rs751917068 Chr.3: 23893312-23893312 on Build GRCh38
rs1012789725 Chr.3: 23893220-23893220 on Build GRCh38
rs780587633 Chr.3: 23893188-23893188 on Build GRCh38
rs761296639 Chr.3: 23893286-23893286 on Build GRCh38
rs1042932578 Chr.3: 23893327-23893327 on Build GRCh38
rs200153520 Chr.3: 23893345-23893345 on Build GRCh38
rs779880356 Chr.3: 23893349-23893349 on Build GRCh38
rs765041356 Chr.3: 23893156-23893156 on Build GRCh38
rs750046063 Chr.3: 23893290-23893290 on Build GRCh38
rs992542157 Chr.3: 23893294-23893294 on Build GRCh38
rs148871674 Chr.3: 23893219-23893219 on Build GRCh38
rs758769690 Chr.3: 23893163-23893163 on Build GRCh38
rs915544668 Chr.3: 23893297-23893297 on Build GRCh38
rs761465500 Chr.3: 23893280-23893280 on Build GRCh38
rs3762775 Chr.3: 23893281-23893281 on Build GRCh38
rs771809065 Chr.3: 23893237-23893237 on Build GRCh38
rs539918987 Chr.3: 23893329-23893329 on Build GRCh38
rs749530012 Chr.3: 23893230-23893230 on Build GRCh38
rs777901887 Chr.3: 23893225-23893225 on Build GRCh38
rs1047105527 Chr.3: 23893146-23893146 on Build GRCh38
rs777856443 Chr.3: 23893334-23893334 on Build GRCh38
rs781613393 Chr.3: 23893328-23893328 on Build GRCh38
rs766933003 Chr.3: 23893324-23893326 on Build GRCh38
rs553532352 Chr.3: 23893147-23893147 on Build GRCh38
rs61751591 Chr.3: 23893314-23893314 on Build GRCh38
rs774330480 Chr.3: 23893186-23893186 on Build GRCh38
rs756125231 Chr.3: 23893330-23893330 on Build GRCh38

View Full Product Details