Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13597399-13597672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF18
Entrez Gene ID
8559
Gene Name
pre-mRNA processing factor 18
Gene Aliases
PRP18, hPrp18
UniGene
Hs.660649
Gene Chromosome Location
Chr.10: 13586811-13645279 on Build GRCh38
Transcripts
NM_003675.3 XM_011519754.2 XM_017016860.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5798610 c.83T>G p.F28C Chr.10: 13597474-13597474 on Build GRCh38
4012954 c.100G>A p.A34T Chr.10: 13597491-13597491 on Build GRCh38
3806824 c.137G>A p.G46D Chr.10: 13597528-13597528 on Build GRCh38
5007038 c.144+2T>G p.? Chr.10: 13597537-13597537 on Build GRCh38
4790552 c.118G>A p.A40T Chr.10: 13597509-13597509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773278708 Chr.10: 13597533-13597533 on Build GRCh38
rs370289910 Chr.10: 13597478-13597478 on Build GRCh38
rs750585303 Chr.10: 13597528-13597528 on Build GRCh38
rs1016729293 Chr.10: 13597635-13597635 on Build GRCh38
rs2502213 Chr.10: 13597490-13597490 on Build GRCh38
rs1040735903 Chr.10: 13597524-13597524 on Build GRCh38
rs377100328 Chr.10: 13597506-13597506 on Build GRCh38
rs747032389 Chr.10: 13597640-13597640 on Build GRCh38
rs974779237 Chr.10: 13597441-13597441 on Build GRCh38
rs530162131 Chr.10: 13597634-13597634 on Build GRCh38
rs201011132 Chr.10: 13597480-13597480 on Build GRCh38
rs756574234 Chr.10: 13597491-13597491 on Build GRCh38
rs377591129 Chr.10: 13597462-13597462 on Build GRCh38
rs895709867 Chr.10: 13597584-13597584 on Build GRCh38
rs777431293 Chr.10: 13597488-13597488 on Build GRCh38
rs780392234 Chr.10: 13597608-13597608 on Build GRCh38
rs759208291 Chr.10: 13597540-13597540 on Build GRCh38
rs760871655 Chr.10: 13597535-13597535 on Build GRCh38
rs996558242 Chr.10: 13597645-13597645 on Build GRCh38
rs752335619 Chr.10: 13597548-13597548 on Build GRCh38
rs755279139 Chr.10: 13597612-13597612 on Build GRCh38
rs980305169 Chr.10: 13597453-13597453 on Build GRCh38
rs936961492 Chr.10: 13597549-13597549 on Build GRCh38
rs766410054 Chr.10: 13597429-13597429 on Build GRCh38
rs528286982 Chr.10: 13597509-13597509 on Build GRCh38
rs369847783 Chr.10: 13597432-13597432 on Build GRCh38
rs548408215 Chr.10: 13597510-13597510 on Build GRCh38
rs367862712 Chr.10: 13597564-13597564 on Build GRCh38
rs141866186 Chr.10: 13597494-13597494 on Build GRCh38
rs191627472 Chr.10: 13597568-13597568 on Build GRCh38
rs754392906 Chr.10: 13597610-13597610 on Build GRCh38
rs762696427 Chr.10: 13597553-13597553 on Build GRCh38
rs145928720 Chr.10: 13597479-13597479 on Build GRCh38
rs748383137 Chr.10: 13597532-13597532 on Build GRCh38
rs748579351 Chr.10: 13597624-13597624 on Build GRCh38
rs770962965 Chr.10: 13597458-13597459 on Build GRCh38
rs780759479 Chr.10: 13597611-13597611 on Build GRCh38
rs776673926 Chr.10: 13597538-13597538 on Build GRCh38
rs774510327 Chr.10: 13597530-13597530 on Build GRCh38
rs371304849 Chr.10: 13597639-13597639 on Build GRCh38
rs766206839 Chr.10: 13597537-13597537 on Build GRCh38
rs756838578 Chr.10: 13597572-13597572 on Build GRCh38
rs375598801 Chr.10: 13597594-13597594 on Build GRCh38
rs368261314 Chr.10: 13597616-13597616 on Build GRCh38
rs765143465 Chr.10: 13597547-13597547 on Build GRCh38

View Full Product Details