Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121793183-121793454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOF
Entrez Gene ID
54509
Gene Name
ras homolog family member F, filopodia associated
Gene Aliases
ARHF, RIF
UniGene
Hs.707579
Gene Chromosome Location
Chr.12: 121777754-121793688 on Build GRCh38
Transcripts
NM_019034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6000846 c.139C>A p.H47N Chr.12: 121793239-121793239 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs913055643 Chr.12: 121793234-121793234 on Build GRCh38
rs1024081959 Chr.12: 121793339-121793339 on Build GRCh38
rs1000363191 Chr.12: 121793293-121793293 on Build GRCh38
rs753156820 Chr.12: 121793243-121793243 on Build GRCh38
rs1042266983 Chr.12: 121793225-121793225 on Build GRCh38
rs886607489 Chr.12: 121793345-121793345 on Build GRCh38
rs975294020 Chr.12: 121793354-121793354 on Build GRCh38
rs866780494 Chr.12: 121793275-121793275 on Build GRCh38
rs545046391 Chr.12: 121793360-121793360 on Build GRCh38
rs768028609 Chr.12: 121793210-121793210 on Build GRCh38
rs750239440 Chr.12: 121793257-121793257 on Build GRCh38
rs988737599 Chr.12: 121793227-121793227 on Build GRCh38
rs1017156869 Chr.12: 121793343-121793343 on Build GRCh38
rs559445038 Chr.12: 121793313-121793313 on Build GRCh38
rs1033982370 Chr.12: 121793295-121793295 on Build GRCh38
rs532875831 Chr.12: 121793341-121793341 on Build GRCh38
rs575653547 Chr.12: 121793280-121793281 on Build GRCh38
rs775591084 Chr.12: 121793427-121793427 on Build GRCh38
rs201021224 Chr.12: 121793277-121793277 on Build GRCh38
rs982650682 Chr.12: 121793342-121793342 on Build GRCh38
rs530694396 Chr.12: 121793436-121793436 on Build GRCh38
rs762387430 Chr.12: 121793207-121793207 on Build GRCh38
rs113086400 Chr.12: 121793303-121793303 on Build GRCh38
rs962622828 Chr.12: 121793351-121793351 on Build GRCh38
rs1013872382 Chr.12: 121793308-121793308 on Build GRCh38
rs921177092 Chr.12: 121793429-121793429 on Build GRCh38
rs942472344 Chr.12: 121793333-121793333 on Build GRCh38
rs563473250 Chr.12: 121793366-121793366 on Build GRCh38
rs553640548 Chr.12: 121793337-121793338 on Build GRCh38
rs1024486309 Chr.12: 121793416-121793416 on Build GRCh38

View Full Product Details