Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169100862-169101087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MECOM
Entrez Gene ID
2122
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
UniGene
Hs.744090
Gene Chromosome Location
Chr.3: 169083499-169663781 on Build GRCh38
Transcripts
NM_001105077.3 NM_001105078.3 NM_001163999.1 NM_001164000.1 NM_001205194.1 NM_004991.3 NM_005241.3 XM_005247213.3 XM_005247214.3 XM_005247215.3 XM_005247219.2 XM_005247220.2 XM_005247221.2 XM_005247223.2 XM_005247224.3 XM_005247225.3 XM_005247226.3 XM_011512546.2 XM_017005874.1 XM_017005875.1 XM_017005876.1 XM_017005877.1 XM_017005878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1040964 c.2232G>A p.R744R Chr.3: 169100938-169100938 on Build GRCh38
4517118 c.2249_2250GG>AA p.R750Q Chr.3: 169100920-169100921 on Build GRCh38
4517117 c.2444_2445GG>AA p.R815Q Chr.3: 169100920-169100921 on Build GRCh38
1286136 c.2285G>T p.R762I Chr.3: 169100885-169100885 on Build GRCh38
6323378 c.2786G>A p.R929Q Chr.3: 169100921-169100921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779796627 Chr.3: 169100989-169100990 on Build GRCh38
rs1004990133 Chr.3: 169101020-169101020 on Build GRCh38
rs864309723 Chr.3: 169100918-169100918 on Build GRCh38
rs377115788 Chr.3: 169100978-169100978 on Build GRCh38
rs767244139 Chr.3: 169101021-169101021 on Build GRCh38
rs369137469 Chr.3: 169100986-169100986 on Build GRCh38
rs745764772 Chr.3: 169100985-169100985 on Build GRCh38
rs864309722 Chr.3: 169100904-169100904 on Build GRCh38
rs775125327 Chr.3: 169100992-169100992 on Build GRCh38
rs139663188 Chr.3: 169101045-169101045 on Build GRCh38
rs115241051 Chr.3: 169101039-169101039 on Build GRCh38
rs879260978 Chr.3: 169100990-169100990 on Build GRCh38
rs781216304 Chr.3: 169100950-169100950 on Build GRCh38
rs864309724 Chr.3: 169100922-169100922 on Build GRCh38
rs1045056359 Chr.3: 169100914-169100914 on Build GRCh38
rs992914520 Chr.3: 169101062-169101062 on Build GRCh38
rs994971015 Chr.3: 169100975-169100975 on Build GRCh38
rs756950116 Chr.3: 169100970-169100970 on Build GRCh38
rs372921435 Chr.3: 169100969-169100969 on Build GRCh38
rs950915127 Chr.3: 169101016-169101016 on Build GRCh38
rs755182939 Chr.3: 169100913-169100913 on Build GRCh38

View Full Product Details