Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95511808-95512069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METAP2
Entrez Gene ID
10988
Gene Name
methionyl aminopeptidase 2
Gene Aliases
MAP2, MNPEP, p67eIF2
UniGene
Hs.444986
Gene Chromosome Location
Chr.12: 95474046-95515839 on Build GRCh38
Transcripts
NM_001317182.1 NM_001317183.1 NM_006838.3 NR_133673.1 XM_005268583.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4932884 c.972A>C p.P324P Chr.12: 95511902-95511902 on Build GRCh38
6711563 c.1050G>A p.G350G Chr.12: 95511980-95511980 on Build GRCh38
432019 c.967A>T p.K323* Chr.12: 95511897-95511897 on Build GRCh38
1364972 c.995C>A p.S332* Chr.12: 95511925-95511925 on Build GRCh38
944665 c.990A>G p.G330G Chr.12: 95511920-95511920 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372495224 Chr.12: 95511836-95511836 on Build GRCh38
rs760072427 Chr.12: 95512022-95512022 on Build GRCh38
rs867365535 Chr.12: 95511877-95511877 on Build GRCh38
rs200039382 Chr.12: 95511903-95511903 on Build GRCh38
rs200464718 Chr.12: 95511851-95511851 on Build GRCh38
rs1016563523 Chr.12: 95511905-95511905 on Build GRCh38
rs753635702 Chr.12: 95511914-95511914 on Build GRCh38
rs200033822 Chr.12: 95512001-95512001 on Build GRCh38
rs1025454491 Chr.12: 95511971-95511971 on Build GRCh38
rs373419277 Chr.12: 95511965-95511965 on Build GRCh38
rs199862503 Chr.12: 95511845-95511845 on Build GRCh38
rs150494953 Chr.12: 95511907-95511907 on Build GRCh38
rs202240016 Chr.12: 95512046-95512046 on Build GRCh38
rs919419000 Chr.12: 95512034-95512034 on Build GRCh38
rs200984122 Chr.12: 95511938-95511938 on Build GRCh38
rs746527409 Chr.12: 95512027-95512027 on Build GRCh38
rs766633383 Chr.12: 95511860-95511860 on Build GRCh38
rs202107367 Chr.12: 95511974-95511974 on Build GRCh38
rs201702084 Chr.12: 95511953-95511953 on Build GRCh38
rs778521665 Chr.12: 95511969-95511969 on Build GRCh38
rs916195909 Chr.12: 95511934-95511934 on Build GRCh38
rs571495863 Chr.12: 95512005-95512005 on Build GRCh38
rs1802823 Chr.12: 95511973-95511973 on Build GRCh38
rs760611381 Chr.12: 95511889-95511889 on Build GRCh38
rs201454366 Chr.12: 95511882-95511882 on Build GRCh38
rs757091265 Chr.12: 95511920-95511920 on Build GRCh38
rs201124733 Chr.12: 95512035-95512035 on Build GRCh38

View Full Product Details