Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200408972-200409237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF281
Entrez Gene ID
23528
Gene Name
zinc finger protein 281
Gene Aliases
ZBP-99, ZNP-99
UniGene
Hs.59757
Gene Chromosome Location
Chr.1: 200404947-200483604 on Build GRCh38
Transcripts
NM_001281293.1 NM_001281294.1 NM_012482.4 XM_017000887.1 XM_017000888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
901930 c.506G>T p.G169V Chr.1: 200409200-200409200 on Build GRCh38
6268471 c.574C>A p.H192N Chr.1: 200409132-200409132 on Build GRCh38
901929 c.583G>A p.V195I Chr.1: 200409123-200409123 on Build GRCh38
6009708 c.489A>C p.L163L Chr.1: 200409217-200409217 on Build GRCh38
424973 c.611A>T p.D204V Chr.1: 200409095-200409095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372746914 Chr.1: 200409199-200409199 on Build GRCh38
rs753580430 Chr.1: 200409126-200409126 on Build GRCh38
rs375813760 Chr.1: 200409090-200409090 on Build GRCh38
rs771169240 Chr.1: 200409055-200409055 on Build GRCh38
rs750266756 Chr.1: 200409108-200409108 on Build GRCh38
rs775654200 Chr.1: 200409192-200409192 on Build GRCh38
rs1035741213 Chr.1: 200409169-200409169 on Build GRCh38
rs182073571 Chr.1: 200409145-200409145 on Build GRCh38
rs771332819 Chr.1: 200409164-200409164 on Build GRCh38
rs748750461 Chr.1: 200409027-200409027 on Build GRCh38
rs201399670 Chr.1: 200409216-200409216 on Build GRCh38
rs769694240 Chr.1: 200409191-200409191 on Build GRCh38
rs367822319 Chr.1: 200409092-200409092 on Build GRCh38
rs761813082 Chr.1: 200409089-200409089 on Build GRCh38
rs202154573 Chr.1: 200409033-200409033 on Build GRCh38
rs748539087 Chr.1: 200409144-200409144 on Build GRCh38
rs370847093 Chr.1: 200409053-200409053 on Build GRCh38
rs186608122 Chr.1: 200409184-200409184 on Build GRCh38
rs778555568 Chr.1: 200409143-200409143 on Build GRCh38
rs751446028 Chr.1: 200409086-200409086 on Build GRCh38
rs148334268 Chr.1: 200409113-200409113 on Build GRCh38
rs141060939 Chr.1: 200409073-200409073 on Build GRCh38
rs555505902 Chr.1: 200409158-200409158 on Build GRCh38
rs765415988 Chr.1: 200409082-200409082 on Build GRCh38
rs779822758 Chr.1: 200409112-200409112 on Build GRCh38
rs773577354 Chr.1: 200409045-200409045 on Build GRCh38
rs776820060 Chr.1: 200409171-200409171 on Build GRCh38
rs776874047 Chr.1: 200409067-200409067 on Build GRCh38
rs902768085 Chr.1: 200409189-200409189 on Build GRCh38
rs754436527 Chr.1: 200409127-200409127 on Build GRCh38
rs140423592 Chr.1: 200409062-200409062 on Build GRCh38
rs755788963 Chr.1: 200409109-200409109 on Build GRCh38
rs1055239810 Chr.1: 200409060-200409060 on Build GRCh38
rs958570708 Chr.1: 200409180-200409180 on Build GRCh38
rs931376544 Chr.1: 200409049-200409049 on Build GRCh38
rs916564693 Chr.1: 200409085-200409085 on Build GRCh38
rs750426053 Chr.1: 200409203-200409203 on Build GRCh38
rs967366640 Chr.1: 200409137-200409137 on Build GRCh38
rs574013915 Chr.1: 200409194-200409194 on Build GRCh38
rs1002990498 Chr.1: 200409156-200409156 on Build GRCh38

View Full Product Details