Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102807502-102807775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIVM
Entrez Gene ID
54841
Gene Name
basic, immunoglobulin-like variable motif containing
Gene Aliases
-
UniGene
Hs.288809
Gene Chromosome Location
Chr.13: 102799049-102841538 on Build GRCh38
Transcripts
NM_001159596.1 NM_017693.3 NM_001204425.1

Gene Symbol
BIVM-ERCC5
Entrez Gene ID
100533467
Gene Name
BIVM-ERCC5 readthrough
Gene Aliases
ERCC5-202
UniGene
Hs.258429
Gene Chromosome Location
Chr.13: 102807146-102876001 on Build GRCh38
Transcripts
NM_001159596.1 NM_017693.3 NM_001204425.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4045782 c.269G>A p.R90H Chr.13: 102807536-102807536 on Build GRCh38
4045783 c.310A>T p.N104Y Chr.13: 102807577-102807577 on Build GRCh38
336442 c.262C>G p.P88A Chr.13: 102807529-102807529 on Build GRCh38
4833751 c.430G>C p.D144H Chr.13: 102807697-102807697 on Build GRCh38
5698786 c.396C>T p.N132N Chr.13: 102807663-102807663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902373021 Chr.13: 102807638-102807639 on Build GRCh38
rs751988877 Chr.13: 102807551-102807551 on Build GRCh38
rs374998551 Chr.13: 102807685-102807685 on Build GRCh38
rs778711423 Chr.13: 102807596-102807596 on Build GRCh38
rs776520067 Chr.13: 102807617-102807617 on Build GRCh38
rs750633383 Chr.13: 102807631-102807631 on Build GRCh38
rs1051492690 Chr.13: 102807623-102807623 on Build GRCh38
rs775400780 Chr.13: 102807606-102807606 on Build GRCh38
rs145982160 Chr.13: 102807536-102807536 on Build GRCh38
rs764054526 Chr.13: 102807529-102807529 on Build GRCh38
rs199598039 Chr.13: 102807608-102807608 on Build GRCh38
rs148446668 Chr.13: 102807732-102807732 on Build GRCh38
rs777107526 Chr.13: 102807575-102807575 on Build GRCh38
rs1038722578 Chr.13: 102807589-102807589 on Build GRCh38
rs748829590 Chr.13: 102807591-102807591 on Build GRCh38
rs770649260 Chr.13: 102807595-102807595 on Build GRCh38
rs149680801 Chr.13: 102807561-102807561 on Build GRCh38
rs139521375 Chr.13: 102807554-102807554 on Build GRCh38
rs745490643 Chr.13: 102807745-102807745 on Build GRCh38
rs142345845 Chr.13: 102807535-102807535 on Build GRCh38
rs948206769 Chr.13: 102807549-102807549 on Build GRCh38
rs778621504 Chr.13: 102807742-102807742 on Build GRCh38
rs745580789 Chr.13: 102807604-102807604 on Build GRCh38
rs780527541 Chr.13: 102807541-102807541 on Build GRCh38
rs765370746 Chr.13: 102807619-102807619 on Build GRCh38
rs779670434 Chr.13: 102807751-102807751 on Build GRCh38
rs766622390 Chr.13: 102807652-102807652 on Build GRCh38
rs754256749 Chr.13: 102807582-102807582 on Build GRCh38
rs771793466 Chr.13: 102807746-102807746 on Build GRCh38
rs768665240 Chr.13: 102807614-102807614 on Build GRCh38
rs370681887 Chr.13: 102807605-102807605 on Build GRCh38
rs1006720248 Chr.13: 102807748-102807748 on Build GRCh38
rs763213181 Chr.13: 102807634-102807634 on Build GRCh38
rs200906938 Chr.13: 102807531-102807531 on Build GRCh38
rs111947510 Chr.13: 102807669-102807669 on Build GRCh38
rs1022104473 Chr.13: 102807752-102807752 on Build GRCh38
rs753330658 Chr.13: 102807731-102807731 on Build GRCh38
rs755391680 Chr.13: 102807680-102807680 on Build GRCh38
rs758760742 Chr.13: 102807539-102807539 on Build GRCh38
rs997280970 Chr.13: 102807613-102807613 on Build GRCh38
rs761890081 Chr.13: 102807618-102807618 on Build GRCh38
rs189801284 Chr.13: 102807638-102807638 on Build GRCh38

View Full Product Details