Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6479123-6479386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP2
Entrez Gene ID
23647
Gene Name
ADP ribosylation factor interacting protein 2
Gene Aliases
POR1
UniGene
Hs.732203
Gene Chromosome Location
Chr.11: 6474683-6481479 on Build GRCh38
Transcripts
NM_001242854.1 NM_001242855.1 NM_001242856.1 NM_012402.3 XM_005252840.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2165332 c.221G>A p.S74N Chr.11: 6479234-6479234 on Build GRCh38
1675798 c.311A>G p.Y104C Chr.11: 6479144-6479144 on Build GRCh38
6423412 c.203G>A p.G68D Chr.11: 6479252-6479252 on Build GRCh38
3368498 c.217C>T p.H73Y Chr.11: 6479238-6479238 on Build GRCh38
6036903 c.315G>A p.K105K Chr.11: 6479140-6479140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567557750 Chr.11: 6479337-6479337 on Build GRCh38
rs766384745 Chr.11: 6479221-6479223 on Build GRCh38
rs551813312 Chr.11: 6479170-6479170 on Build GRCh38
rs974558658 Chr.11: 6479321-6479321 on Build GRCh38
rs756362468 Chr.11: 6479271-6479271 on Build GRCh38
rs139083273 Chr.11: 6479278-6479278 on Build GRCh38
rs746366086 Chr.11: 6479279-6479279 on Build GRCh38
rs143197439 Chr.11: 6479226-6479226 on Build GRCh38
rs1021520445 Chr.11: 6479344-6479344 on Build GRCh38
rs764932808 Chr.11: 6479231-6479231 on Build GRCh38
rs766340094 Chr.11: 6479333-6479333 on Build GRCh38
rs778012688 Chr.11: 6479276-6479276 on Build GRCh38
rs779160718 Chr.11: 6479363-6479363 on Build GRCh38
rs749615881 Chr.11: 6479173-6479173 on Build GRCh38
rs745958248 Chr.11: 6479343-6479343 on Build GRCh38
rs769988786 Chr.11: 6479169-6479169 on Build GRCh38
rs781405866 Chr.11: 6479285-6479285 on Build GRCh38
rs772575644 Chr.11: 6479281-6479281 on Build GRCh38
rs763578402 Chr.11: 6479224-6479224 on Build GRCh38
rs890085753 Chr.11: 6479208-6479208 on Build GRCh38
rs781134013 Chr.11: 6479250-6479250 on Build GRCh38
rs754126797 Chr.11: 6479360-6479360 on Build GRCh38
rs777821545 Chr.11: 6479351-6479351 on Build GRCh38
rs756239714 Chr.11: 6479350-6479350 on Build GRCh38
rs749493573 Chr.11: 6479277-6479277 on Build GRCh38
rs923204774 Chr.11: 6479323-6479323 on Build GRCh38
rs751298057 Chr.11: 6479336-6479336 on Build GRCh38
rs549041467 Chr.11: 6479329-6479329 on Build GRCh38
rs929026817 Chr.11: 6479198-6479198 on Build GRCh38
rs773047944 Chr.11: 6479312-6479312 on Build GRCh38
rs538083961 Chr.11: 6479349-6479349 on Build GRCh38
rs781315763 Chr.11: 6479165-6479165 on Build GRCh38
rs758308260 Chr.11: 6479242-6479242 on Build GRCh38
rs761386615 Chr.11: 6479230-6479230 on Build GRCh38
rs372087765 Chr.11: 6479244-6479244 on Build GRCh38
rs771258941 Chr.11: 6479199-6479199 on Build GRCh38
rs757403941 Chr.11: 6479362-6479362 on Build GRCh38
rs748198428 Chr.11: 6479264-6479264 on Build GRCh38
rs760075782 Chr.11: 6479215-6479215 on Build GRCh38
rs750272738 Chr.11: 6479234-6479234 on Build GRCh38
rs541465939 Chr.11: 6479353-6479353 on Build GRCh38
rs748383550 Chr.11: 6479167-6479167 on Build GRCh38
rs188872704 Chr.11: 6479304-6479304 on Build GRCh38
rs754935012 Chr.11: 6479249-6479249 on Build GRCh38

View Full Product Details