Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244378426-244378683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf100
Entrez Gene ID
200159
Gene Name
chromosome 1 open reading frame 100
Gene Aliases
-
UniGene
Hs.442703
Gene Chromosome Location
Chr.1: 244352063-244389896 on Build GRCh38
Transcripts
NM_001012970.2 NM_001276348.1 NM_001276349.1 XM_011544124.2 XM_011544125.2 XM_011544128.2 XM_011544129.2 XM_011544130.2 XM_017000571.1 XM_017000572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
906665 c.244C>T p.R82* Chr.1: 244378558-244378558 on Build GRCh38
6315617 c.239G>T p.C80F Chr.1: 244378553-244378553 on Build GRCh38
4030330 c.241C>A p.L81I Chr.1: 244378555-244378555 on Build GRCh38
274232 c.293G>A p.R98Q Chr.1: 244378607-244378607 on Build GRCh38
5911215 c.155C>T p.P52L Chr.1: 244378469-244378469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757315523 Chr.1: 244378587-244378587 on Build GRCh38
rs759151667 Chr.1: 244378652-244378652 on Build GRCh38
rs200395444 Chr.1: 244378544-244378544 on Build GRCh38
rs370385695 Chr.1: 244378653-244378653 on Build GRCh38
rs186873318 Chr.1: 244378461-244378461 on Build GRCh38
rs762477027 Chr.1: 244378559-244378559 on Build GRCh38
rs756609897 Chr.1: 244378491-244378491 on Build GRCh38
rs144659720 Chr.1: 244378471-244378471 on Build GRCh38
rs41269385 Chr.1: 244378525-244378525 on Build GRCh38
rs779159614 Chr.1: 244378511-244378511 on Build GRCh38
rs191768495 Chr.1: 244378468-244378468 on Build GRCh38
rs778313104 Chr.1: 244378493-244378493 on Build GRCh38
rs374751783 Chr.1: 244378599-244378599 on Build GRCh38
rs761807907 Chr.1: 244378467-244378467 on Build GRCh38
rs202173669 Chr.1: 244378470-244378470 on Build GRCh38
rs770949540 Chr.1: 244378641-244378641 on Build GRCh38
rs906645188 Chr.1: 244378607-244378607 on Build GRCh38
rs767396280 Chr.1: 244378644-244378644 on Build GRCh38
rs1003856114 Chr.1: 244378518-244378518 on Build GRCh38
rs756472769 Chr.1: 244378580-244378580 on Build GRCh38
rs781388686 Chr.1: 244378482-244378482 on Build GRCh38
rs754361787 Chr.1: 244378584-244378584 on Build GRCh38
rs772390494 Chr.1: 244378522-244378522 on Build GRCh38
rs749310237 Chr.1: 244378498-244378498 on Build GRCh38
rs778841110 Chr.1: 244378592-244378592 on Build GRCh38
rs755321248 Chr.1: 244378479-244378479 on Build GRCh38
rs749149291 Chr.1: 244378637-244378637 on Build GRCh38
rs144063808 Chr.1: 244378606-244378606 on Build GRCh38
rs376583429 Chr.1: 244378579-244378579 on Build GRCh38
rs769734875 Chr.1: 244378616-244378616 on Build GRCh38
rs41269383 Chr.1: 244378469-244378469 on Build GRCh38
rs970898199 Chr.1: 244378597-244378597 on Build GRCh38
rs747895243 Chr.1: 244378609-244378609 on Build GRCh38
rs772247057 Chr.1: 244378454-244378457 on Build GRCh38
rs761769369 Chr.1: 244378543-244378543 on Build GRCh38
rs773244467 Chr.1: 244378619-244378619 on Build GRCh38
rs267598455 Chr.1: 244378558-244378558 on Build GRCh38
rs753101608 Chr.1: 244378489-244378489 on Build GRCh38
rs751120608 Chr.1: 244378578-244378578 on Build GRCh38
rs770924077 Chr.1: 244378501-244378501 on Build GRCh38
rs778192628 Chr.1: 244378581-244378581 on Build GRCh38
rs746104121 Chr.1: 244378521-244378521 on Build GRCh38
rs138539876 Chr.1: 244378475-244378475 on Build GRCh38
rs746021366 Chr.1: 244378594-244378594 on Build GRCh38

View Full Product Details