Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74544381-74544614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3B
Entrez Gene ID
10620
Gene Name
AT-rich interaction domain 3B
Gene Aliases
BDP, DRIL2
UniGene
Hs.647642
Gene Chromosome Location
Chr.15: 74541177-74598131 on Build GRCh38
Transcripts
NM_001307939.1 NM_006465.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3794399 c.506C>T p.S169F Chr.15: 74544442-74544442 on Build GRCh38
3794400 c.515G>C p.G172A Chr.15: 74544451-74544451 on Build GRCh38
6604655 c.506C>G p.S169C Chr.15: 74544442-74544442 on Build GRCh38
3503939 c.504C>T p.V168V Chr.15: 74544440-74544440 on Build GRCh38
4768557 c.552G>T p.Q184H Chr.15: 74544488-74544488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773326406 Chr.15: 74544435-74544435 on Build GRCh38
rs760670903 Chr.15: 74544436-74544436 on Build GRCh38
rs758719943 Chr.15: 74544458-74544458 on Build GRCh38
rs758760390 Chr.15: 74544554-74544554 on Build GRCh38
rs761652462 Chr.15: 74544433-74544433 on Build GRCh38
rs572861926 Chr.15: 74544481-74544481 on Build GRCh38
rs779496302 Chr.15: 74544502-74544502 on Build GRCh38
rs894458898 Chr.15: 74544518-74544518 on Build GRCh38
rs144164282 Chr.15: 74544568-74544568 on Build GRCh38
rs774032036 Chr.15: 74544432-74544432 on Build GRCh38
rs768215998 Chr.15: 74544506-74544506 on Build GRCh38
rs554192034 Chr.15: 74544474-74544474 on Build GRCh38
rs894909137 Chr.15: 74544542-74544542 on Build GRCh38
rs374850745 Chr.15: 74544411-74544411 on Build GRCh38
rs1013251356 Chr.15: 74544546-74544546 on Build GRCh38
rs769333131 Chr.15: 74544409-74544409 on Build GRCh38
rs573623190 Chr.15: 74544576-74544576 on Build GRCh38
rs755156759 Chr.15: 74544448-74544448 on Build GRCh38
rs765438579 Chr.15: 74544452-74544452 on Build GRCh38
rs111490747 Chr.15: 74544557-74544557 on Build GRCh38
rs373097859 Chr.15: 74544505-74544505 on Build GRCh38
rs768348565 Chr.15: 74544425-74544425 on Build GRCh38
rs140786777 Chr.15: 74544438-74544438 on Build GRCh38
rs1014186402 Chr.15: 74544540-74544540 on Build GRCh38
rs748949732 Chr.15: 74544419-74544419 on Build GRCh38
rs753023065 Chr.15: 74544454-74544454 on Build GRCh38
rs369668091 Chr.15: 74544473-74544473 on Build GRCh38
rs774017279 Chr.15: 74544531-74544531 on Build GRCh38

View Full Product Details