Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 33389449-33389676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT10
Entrez Gene ID
84750
Gene Name
fucosyltransferase 10
Gene Aliases
FUCTX
UniGene
Hs.458713
Gene Chromosome Location
Chr.8: 33308102-33473492 on Build GRCh38
Transcripts
NM_032664.3 XM_011544676.2 XM_011544677.2 XM_011544678.2 XM_011544679.2 XM_011544680.2 XM_011544681.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7183685 c.443G>C p.R148T Chr.8: 33389546-33389546 on Build GRCh38
1743396 c.605A>G p.Q202R Chr.8: 33389570-33389570 on Build GRCh38
4996958 c.488C>T p.S163L Chr.8: 33389501-33389501 on Build GRCh38
4438866 c.667C>T p.P223S Chr.8: 33389508-33389508 on Build GRCh38
7368944 c.529G>A p.E177K Chr.8: 33389460-33389460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771837191 Chr.8: 33389619-33389619 on Build GRCh38
rs543964339 Chr.8: 33389642-33389642 on Build GRCh38
rs763631538 Chr.8: 33389541-33389541 on Build GRCh38
rs748140955 Chr.8: 33389532-33389532 on Build GRCh38
rs142881205 Chr.8: 33389634-33389634 on Build GRCh38
rs201235823 Chr.8: 33389588-33389588 on Build GRCh38
rs745724380 Chr.8: 33389616-33389616 on Build GRCh38
rs751291924 Chr.8: 33389489-33389489 on Build GRCh38
rs771401453 Chr.8: 33389612-33389612 on Build GRCh38
rs761537447 Chr.8: 33389577-33389577 on Build GRCh38
rs555345598 Chr.8: 33389481-33389481 on Build GRCh38
rs768416455 Chr.8: 33389638-33389638 on Build GRCh38
rs747442726 Chr.8: 33389554-33389554 on Build GRCh38
rs755398161 Chr.8: 33389604-33389604 on Build GRCh38
rs1028944354 Chr.8: 33389564-33389564 on Build GRCh38
rs1004971118 Chr.8: 33389504-33389504 on Build GRCh38
rs750881715 Chr.8: 33389580-33389580 on Build GRCh38
rs769956591 Chr.8: 33389501-33389501 on Build GRCh38
rs886498819 Chr.8: 33389493-33389493 on Build GRCh38
rs756839426 Chr.8: 33389510-33389510 on Build GRCh38
rs760156413 Chr.8: 33389537-33389537 on Build GRCh38
rs774147172 Chr.8: 33389509-33389509 on Build GRCh38
rs753509956 Chr.8: 33389476-33389476 on Build GRCh38
rs200415562 Chr.8: 33389479-33389479 on Build GRCh38
rs748431671 Chr.8: 33389486-33389486 on Build GRCh38
rs143890078 Chr.8: 33389483-33389483 on Build GRCh38
rs367637252 Chr.8: 33389480-33389480 on Build GRCh38
rs752073044 Chr.8: 33389595-33389595 on Build GRCh38
rs376088705 Chr.8: 33389608-33389608 on Build GRCh38
rs1009239899 Chr.8: 33389589-33389589 on Build GRCh38
rs748120060 Chr.8: 33389611-33389611 on Build GRCh38
rs772976416 Chr.8: 33389652-33389652 on Build GRCh38
rs140205664 Chr.8: 33389478-33389478 on Build GRCh38
rs866041756 Chr.8: 33389594-33389594 on Build GRCh38
rs764912362 Chr.8: 33389579-33389579 on Build GRCh38
rs761399721 Chr.8: 33389641-33389641 on Build GRCh38
rs745601484 Chr.8: 33389609-33389609 on Build GRCh38
rs148547444 Chr.8: 33389612-33389612 on Build GRCh38
rs28727521 Chr.8: 33389536-33389536 on Build GRCh38
rs753235557 Chr.8: 33389547-33389547 on Build GRCh38
rs775331843 Chr.8: 33389627-33389627 on Build GRCh38
rs776207784 Chr.8: 33389640-33389640 on Build GRCh38

View Full Product Details