Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42698875-42699110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HHATL
Entrez Gene ID
57467
Gene Name
hedgehog acyltransferase-like
Gene Aliases
C3orf3, GUP1, MBOAT3, MSTP002, OACT3
UniGene
Hs.476041
Gene Chromosome Location
Chr.3: 42692663-42702849 on Build GRCh38
Transcripts
NM_020707.3 NR_027753.1 XM_006713274.2 XM_006713275.2 XM_011533969.2 XM_011533970.2 XM_017006935.1 XM_017006936.1 XM_017006937.1 XM_017006938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7232101 c.281C>A p.A94D Chr.3: 42699039-42699039 on Build GRCh38
1235677 c.236C>T p.A79V Chr.3: 42699084-42699084 on Build GRCh38
240108 c.218G>A p.R73H Chr.3: 42699102-42699102 on Build GRCh38
1044773 c.217C>T p.R73C Chr.3: 42699103-42699103 on Build GRCh38
1239063 c.293G>A p.R98H Chr.3: 42698898-42698898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946978119 Chr.3: 42698972-42698972 on Build GRCh38
rs748605159 Chr.3: 42698901-42698901 on Build GRCh38
rs775254923 Chr.3: 42699006-42699006 on Build GRCh38
rs771137074 Chr.3: 42698983-42698983 on Build GRCh38
rs61739734 Chr.3: 42699048-42699048 on Build GRCh38
rs768526073 Chr.3: 42699016-42699016 on Build GRCh38
rs746347440 Chr.3: 42699076-42699076 on Build GRCh38
rs770049455 Chr.3: 42698906-42698906 on Build GRCh38
rs768134252 Chr.3: 42699086-42699086 on Build GRCh38
rs759731131 Chr.3: 42699029-42699029 on Build GRCh38
rs773812644 Chr.3: 42698919-42698919 on Build GRCh38
rs765323035 Chr.3: 42699022-42699022 on Build GRCh38
rs1013914639 Chr.3: 42699052-42699052 on Build GRCh38
rs776221058 Chr.3: 42699019-42699019 on Build GRCh38
rs149682326 Chr.3: 42699077-42699077 on Build GRCh38
rs752832993 Chr.3: 42698946-42698946 on Build GRCh38
rs201339392 Chr.3: 42699004-42699004 on Build GRCh38
rs373868901 Chr.3: 42698925-42698925 on Build GRCh38
rs200065074 Chr.3: 42698931-42698931 on Build GRCh38
rs764526572 Chr.3: 42699053-42699053 on Build GRCh38
rs551431803 Chr.3: 42699024-42699024 on Build GRCh38
rs146696390 Chr.3: 42698942-42698942 on Build GRCh38
rs74648342 Chr.3: 42699000-42699000 on Build GRCh38
rs770453576 Chr.3: 42698914-42698914 on Build GRCh38
rs200755991 Chr.3: 42698898-42698898 on Build GRCh38
rs143122687 Chr.3: 42698922-42698922 on Build GRCh38
rs749737704 Chr.3: 42698910-42698910 on Build GRCh38
rs146384822 Chr.3: 42699061-42699061 on Build GRCh38
rs778385970 Chr.3: 42698909-42698909 on Build GRCh38
rs113329939 Chr.3: 42698899-42698899 on Build GRCh38
rs757608632 Chr.3: 42699066-42699066 on Build GRCh38
rs867078388 Chr.3: 42699062-42699062 on Build GRCh38
rs369820688 Chr.3: 42698947-42698947 on Build GRCh38
rs779873818 Chr.3: 42699067-42699068 on Build GRCh38
rs368151427 Chr.3: 42698920-42698920 on Build GRCh38
rs570352319 Chr.3: 42698937-42698937 on Build GRCh38
rs368127987 Chr.3: 42698930-42698930 on Build GRCh38
rs150548791 Chr.3: 42699071-42699071 on Build GRCh38
rs145469527 Chr.3: 42699084-42699084 on Build GRCh38
rs751631169 Chr.3: 42698935-42698935 on Build GRCh38
rs375430754 Chr.3: 42698950-42698950 on Build GRCh38
rs914193257 Chr.3: 42698971-42698971 on Build GRCh38
rs757915927 Chr.3: 42699009-42699010 on Build GRCh38
rs766569959 Chr.3: 42698934-42698934 on Build GRCh38
rs925210012 Chr.3: 42699012-42699012 on Build GRCh38
rs745445051 Chr.3: 42699001-42699001 on Build GRCh38
rs746590837 Chr.3: 42699013-42699013 on Build GRCh38
rs922647902 Chr.3: 42698944-42698944 on Build GRCh38
rs963321789 Chr.3: 42698938-42698938 on Build GRCh38
rs761800096 Chr.3: 42698932-42698932 on Build GRCh38
rs372107414 Chr.3: 42698948-42698948 on Build GRCh38
rs752986852 Chr.3: 42699040-42699040 on Build GRCh38
rs913745816 Chr.3: 42699056-42699056 on Build GRCh38
rs376426134 Chr.3: 42699021-42699021 on Build GRCh38
rs767475391 Chr.3: 42699031-42699031 on Build GRCh38
rs779389328 Chr.3: 42699081-42699081 on Build GRCh38

View Full Product Details