Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209675690-209675919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G0S2
Entrez Gene ID
50486
Gene Name
G0/G1 switch 2
Gene Aliases
-
UniGene
Hs.432132
Gene Chromosome Location
Chr.1: 209675325-209676390 on Build GRCh38
Transcripts
NM_015714.3 NR_134509.1 NR_134510.1

Gene Symbol
LOC101930114
Entrez Gene ID
101930114
Gene Name
uncharacterized LOC101930114
Gene Aliases
-
UniGene
Hs.663934
Gene Chromosome Location
Chr.1: 209661359-209742562 on Build GRCh38
Transcripts
NM_015714.3 NR_134509.1 NR_134510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6251831 c.31G>T p.A11S Chr.1: 209675715-209675715 on Build GRCh38
5744702 c.26C>A p.P9H Chr.1: 209675710-209675710 on Build GRCh38
5531543 c.105C>T p.L35L Chr.1: 209675789-209675789 on Build GRCh38
5440382 c.60delG p.K22fs*3 Chr.1: 209675744-209675744 on Build GRCh38
5440326 c.66G>A p.K22K Chr.1: 209675750-209675750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766635565 Chr.1: 209675869-209675869 on Build GRCh38
rs140491567 Chr.1: 209675868-209675868 on Build GRCh38
rs373742326 Chr.1: 209675796-209675796 on Build GRCh38
rs199631971 Chr.1: 209675792-209675792 on Build GRCh38
rs759472154 Chr.1: 209675837-209675837 on Build GRCh38
rs150720329 Chr.1: 209675871-209675871 on Build GRCh38
rs142125926 Chr.1: 209675858-209675858 on Build GRCh38
rs765796722 Chr.1: 209675887-209675887 on Build GRCh38
rs900396776 Chr.1: 209675784-209675784 on Build GRCh38
rs1017561628 Chr.1: 209675760-209675760 on Build GRCh38
rs899250673 Chr.1: 209675824-209675824 on Build GRCh38
rs775289692 Chr.1: 209675855-209675855 on Build GRCh38
rs780161389 Chr.1: 209675766-209675766 on Build GRCh38
rs370237791 Chr.1: 209675791-209675791 on Build GRCh38
rs752420827 Chr.1: 209675734-209675734 on Build GRCh38
rs745397091 Chr.1: 209675793-209675793 on Build GRCh38
rs982234446 Chr.1: 209675880-209675880 on Build GRCh38
rs955989025 Chr.1: 209675804-209675804 on Build GRCh38
rs778185736 Chr.1: 209675898-209675898 on Build GRCh38
rs769228485 Chr.1: 209675795-209675795 on Build GRCh38
rs753145304 Chr.1: 209675771-209675793 on Build GRCh38
rs750664680 Chr.1: 209675864-209675864 on Build GRCh38
rs770401244 Chr.1: 209675822-209675822 on Build GRCh38
rs988483365 Chr.1: 209675814-209675814 on Build GRCh38
rs765080587 Chr.1: 209675719-209675719 on Build GRCh38
rs143745964 Chr.1: 209675899-209675899 on Build GRCh38
rs753904643 Chr.1: 209675870-209675870 on Build GRCh38
rs147279528 Chr.1: 209675738-209675738 on Build GRCh38
rs753220688 Chr.1: 209675888-209675888 on Build GRCh38
rs769663352 Chr.1: 209675838-209675838 on Build GRCh38
rs762616506 Chr.1: 209675857-209675857 on Build GRCh38
rs780530900 Chr.1: 209675773-209675773 on Build GRCh38
rs1008832034 Chr.1: 209675739-209675739 on Build GRCh38
rs376545220 Chr.1: 209675740-209675740 on Build GRCh38
rs771787297 Chr.1: 209675805-209675805 on Build GRCh38
rs776111464 Chr.1: 209675836-209675836 on Build GRCh38
rs779568938 Chr.1: 209675806-209675806 on Build GRCh38
rs756188171 Chr.1: 209675743-209675743 on Build GRCh38
rs758733987 Chr.1: 209675895-209675895 on Build GRCh38

View Full Product Details