Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 720679-720922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS8L2
Entrez Gene ID
64787
Gene Name
EPS8 like 2
Gene Aliases
EPS8R2
UniGene
Hs.55016
Gene Chromosome Location
Chr.11: 705217-727727 on Build GRCh38
Transcripts
NM_022772.3 XM_017018130.1 XM_017018131.1 XM_017018132.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7295795 c.541C>T p.R181W Chr.11: 720893-720893 on Build GRCh38
6397528 c.515C>T p.A172V Chr.11: 720867-720867 on Build GRCh38
84032 c.521G>C p.C174S Chr.11: 720873-720873 on Build GRCh38
6497221 c.542G>A p.R181Q Chr.11: 720894-720894 on Build GRCh38
5480329 c.519C>A p.D173E Chr.11: 720871-720871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554029920 Chr.11: 720767-720767 on Build GRCh38
rs754127157 Chr.11: 720703-720703 on Build GRCh38
rs6597989 Chr.11: 720763-720763 on Build GRCh38
rs934957546 Chr.11: 720736-720736 on Build GRCh38
rs767466392 Chr.11: 720812-720812 on Build GRCh38
rs539592756 Chr.11: 720868-720868 on Build GRCh38
rs765381407 Chr.11: 720866-720866 on Build GRCh38
rs762570304 Chr.11: 720777-720777 on Build GRCh38
rs372636952 Chr.11: 720713-720713 on Build GRCh38
rs779909774 Chr.11: 720881-720882 on Build GRCh38
rs1039829888 Chr.11: 720782-720782 on Build GRCh38
rs985834266 Chr.11: 720743-720743 on Build GRCh38
rs904185089 Chr.11: 720820-720820 on Build GRCh38
rs1056665536 Chr.11: 720796-720796 on Build GRCh38
rs906189716 Chr.11: 720722-720722 on Build GRCh38
rs973254815 Chr.11: 720897-720897 on Build GRCh38
rs955810683 Chr.11: 720859-720859 on Build GRCh38
rs1012510123 Chr.11: 720807-720807 on Build GRCh38
rs899967190 Chr.11: 720795-720795 on Build GRCh38
rs200177008 Chr.11: 720867-720867 on Build GRCh38
rs750167406 Chr.11: 720819-720819 on Build GRCh38
rs1000302540 Chr.11: 720827-720827 on Build GRCh38
rs976529477 Chr.11: 720860-720860 on Build GRCh38
rs909888133 Chr.11: 720744-720744 on Build GRCh38
rs867054985 Chr.11: 720850-720850 on Build GRCh38
rs757143018 Chr.11: 720697-720697 on Build GRCh38
rs1037113590 Chr.11: 720757-720757 on Build GRCh38
rs758406101 Chr.11: 720753-720753 on Build GRCh38
rs774464810 Chr.11: 720780-720780 on Build GRCh38
rs767652699 Chr.11: 720751-720760 on Build GRCh38
rs895372854 Chr.11: 720798-720798 on Build GRCh38
rs756072469 Chr.11: 720780-720781 on Build GRCh38
rs749057351 Chr.11: 720884-720886 on Build GRCh38
rs748168915 Chr.11: 720735-720735 on Build GRCh38
rs755290745 Chr.11: 720845-720845 on Build GRCh38
rs1039315639 Chr.11: 720769-720769 on Build GRCh38
rs775142661 Chr.11: 720773-720773 on Build GRCh38
rs778266262 Chr.11: 720759-720759 on Build GRCh38
rs767384396 Chr.11: 720702-720702 on Build GRCh38
rs745594494 Chr.11: 720764-720764 on Build GRCh38
rs866382650 Chr.11: 720878-720878 on Build GRCh38
rs750257597 Chr.11: 720777-720778 on Build GRCh38
rs757238153 Chr.11: 720743-720745 on Build GRCh38
rs577356107 Chr.11: 720778-720778 on Build GRCh38
rs747463415 Chr.11: 720760-720760 on Build GRCh38
rs761814265 Chr.11: 720781-720781 on Build GRCh38
rs758493990 Chr.11: 720892-720892 on Build GRCh38
rs963279459 Chr.11: 720895-720895 on Build GRCh38
rs754806041 Chr.11: 720892-720893 on Build GRCh38
rs779103363 Chr.11: 720717-720717 on Build GRCh38
rs1014864959 Chr.11: 720884-720884 on Build GRCh38
rs755130479 Chr.11: 720714-720714 on Build GRCh38

View Full Product Details