Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96965956-96966181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPQ
Entrez Gene ID
10404
Gene Name
carboxypeptidase Q
Gene Aliases
LDP, PGCP
UniGene
Hs.156178
Gene Chromosome Location
Chr.8: 96645227-97143503 on Build GRCh38
Transcripts
NM_016134.3 XM_017012953.1 XM_017012954.1 XM_017012955.1 NR_125390.1

Gene Symbol
LOC101927066
Entrez Gene ID
101927066
Gene Name
uncharacterized LOC101927066
Gene Aliases
-
UniGene
Hs.732112
Gene Chromosome Location
Chr.8: 96951864-97446497 on Build GRCh38
Transcripts
NM_016134.3 XM_017012953.1 XM_017012954.1 XM_017012955.1 NR_125390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6181686 c.882G>T p.W294C Chr.8: 96965967-96965967 on Build GRCh38
6634759 c.889G>T p.G297W Chr.8: 96965974-96965974 on Build GRCh38
4408441 c.899C>T p.A300V Chr.8: 96965984-96965984 on Build GRCh38
6034460 c.911G>A p.G304D Chr.8: 96965996-96965996 on Build GRCh38
3902288 c.937G>A p.A313T Chr.8: 96966022-96966022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369433276 Chr.8: 96965997-96965997 on Build GRCh38
rs769535534 Chr.8: 96966054-96966054 on Build GRCh38
rs570779644 Chr.8: 96966137-96966137 on Build GRCh38
rs550855973 Chr.8: 96966079-96966079 on Build GRCh38
rs374439582 Chr.8: 96966073-96966073 on Build GRCh38
rs748078486 Chr.8: 96966051-96966051 on Build GRCh38
rs779425986 Chr.8: 96966005-96966005 on Build GRCh38
rs765892531 Chr.8: 96966095-96966095 on Build GRCh38
rs968165957 Chr.8: 96965980-96965980 on Build GRCh38
rs184645863 Chr.8: 96966022-96966022 on Build GRCh38
rs764158563 Chr.8: 96965977-96965977 on Build GRCh38
rs765761117 Chr.8: 96965979-96965979 on Build GRCh38
rs113874164 Chr.8: 96966085-96966085 on Build GRCh38
rs773079583 Chr.8: 96966058-96966058 on Build GRCh38
rs754332449 Chr.8: 96966090-96966090 on Build GRCh38
rs781045399 Chr.8: 96966039-96966039 on Build GRCh38
rs373415893 Chr.8: 96965998-96965998 on Build GRCh38
rs201303595 Chr.8: 96966075-96966075 on Build GRCh38
rs76308991 Chr.8: 96966077-96966077 on Build GRCh38
rs762708490 Chr.8: 96966063-96966063 on Build GRCh38
rs917594248 Chr.8: 96966136-96966136 on Build GRCh38
rs751061172 Chr.8: 96965985-96965985 on Build GRCh38
rs757637878 Chr.8: 96966000-96966000 on Build GRCh38
rs372544728 Chr.8: 96966028-96966028 on Build GRCh38
rs376395954 Chr.8: 96966006-96966006 on Build GRCh38
rs201149237 Chr.8: 96966017-96966017 on Build GRCh38
rs376543974 Chr.8: 96966029-96966029 on Build GRCh38
rs750766445 Chr.8: 96966102-96966102 on Build GRCh38
rs990953677 Chr.8: 96966129-96966129 on Build GRCh38
rs776345537 Chr.8: 96966065-96966065 on Build GRCh38
rs939422422 Chr.8: 96966072-96966072 on Build GRCh38
rs771748991 Chr.8: 96965989-96965989 on Build GRCh38

View Full Product Details