Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89713791-89714052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC8C
Entrez Gene ID
84230
Gene Name
leucine rich repeat containing 8 family member C
Gene Aliases
AD158, FAD158
UniGene
Hs.412836
Gene Chromosome Location
Chr.1: 89615810-89719535 on Build GRCh38
Transcripts
NM_032270.4 XM_006710960.3 XM_011542282.2 XM_011542283.2 XM_017002503.1 XM_017002504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5895494 c.1273C>T p.R425* Chr.1: 89713843-89713843 on Build GRCh38
3806074 c.1292T>C p.I431T Chr.1: 89713862-89713862 on Build GRCh38
5464372 c.1358T>G p.I453S Chr.1: 89713928-89713928 on Build GRCh38
336647 c.1221C>A p.N407K Chr.1: 89713791-89713791 on Build GRCh38
682773 c.1420C>T p.L474L Chr.1: 89713990-89713990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183454204 Chr.1: 89713848-89713848 on Build GRCh38
rs770087026 Chr.1: 89714024-89714024 on Build GRCh38
rs750163797 Chr.1: 89713928-89713928 on Build GRCh38
rs747175917 Chr.1: 89713850-89713850 on Build GRCh38
rs766226957 Chr.1: 89713936-89713936 on Build GRCh38
rs749754612 Chr.1: 89714023-89714023 on Build GRCh38
rs755253718 Chr.1: 89713822-89713822 on Build GRCh38
rs202029362 Chr.1: 89713989-89713990 on Build GRCh38
rs747673573 Chr.1: 89714026-89714026 on Build GRCh38
rs1009644796 Chr.1: 89714005-89714005 on Build GRCh38
rs781093353 Chr.1: 89713923-89713923 on Build GRCh38
rs758573976 Chr.1: 89713844-89713844 on Build GRCh38
rs34576461 Chr.1: 89713911-89713911 on Build GRCh38
rs764225757 Chr.1: 89713939-89713939 on Build GRCh38
rs780402524 Chr.1: 89714010-89714010 on Build GRCh38
rs188344379 Chr.1: 89713897-89713897 on Build GRCh38
rs780655847 Chr.1: 89713889-89713889 on Build GRCh38
rs780056387 Chr.1: 89713838-89713838 on Build GRCh38
rs781633185 Chr.1: 89713867-89713867 on Build GRCh38
rs773873869 Chr.1: 89713915-89713915 on Build GRCh38
rs775557961 Chr.1: 89713861-89713863 on Build GRCh38
rs139410920 Chr.1: 89714025-89714025 on Build GRCh38
rs752708379 Chr.1: 89713839-89713839 on Build GRCh38
rs940215975 Chr.1: 89714022-89714022 on Build GRCh38
rs866698768 Chr.1: 89713929-89713929 on Build GRCh38
rs757494121 Chr.1: 89713947-89713947 on Build GRCh38
rs753858417 Chr.1: 89713938-89713938 on Build GRCh38
rs897481928 Chr.1: 89713856-89713856 on Build GRCh38
rs746254719 Chr.1: 89713869-89713869 on Build GRCh38
rs762338886 Chr.1: 89713904-89713904 on Build GRCh38
rs371950033 Chr.1: 89713997-89713997 on Build GRCh38
rs12032393 Chr.1: 89713973-89713973 on Build GRCh38
rs767163360 Chr.1: 89713927-89713927 on Build GRCh38
rs147275787 Chr.1: 89713861-89713861 on Build GRCh38
rs968012904 Chr.1: 89713891-89713891 on Build GRCh38
rs139178832 Chr.1: 89713970-89713970 on Build GRCh38
rs866269535 Chr.1: 89713843-89713843 on Build GRCh38
rs1049286212 Chr.1: 89713877-89713877 on Build GRCh38
rs149529211 Chr.1: 89713885-89713885 on Build GRCh38
rs760580392 Chr.1: 89713931-89713931 on Build GRCh38

View Full Product Details