Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81362436-81362705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAN
Entrez Gene ID
8139
Gene Name
gigaxonin
Gene Aliases
GAN1, KLHL16
UniGene
Hs.112569
Gene Chromosome Location
Chr.16: 81314941-81380198 on Build GRCh38
Transcripts
NM_022041.3 XM_017023734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5429429 c.1084G>A p.E362K Chr.16: 81362609-81362609 on Build GRCh38
1290598 c.1021C>G p.L341V Chr.16: 81362546-81362546 on Build GRCh38
3717100 c.977G>T p.G326V Chr.16: 81362502-81362502 on Build GRCh38
4568200 c.1022T>G p.L341R Chr.16: 81362547-81362547 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772181617 Chr.16: 81362496-81362496 on Build GRCh38
rs368906326 Chr.16: 81362628-81362628 on Build GRCh38
rs12448327 Chr.16: 81362682-81362682 on Build GRCh38
rs377539440 Chr.16: 81362589-81362589 on Build GRCh38
rs771038275 Chr.16: 81362605-81362605 on Build GRCh38
rs201020426 Chr.16: 81362634-81362634 on Build GRCh38
rs547033474 Chr.16: 81362477-81362477 on Build GRCh38
rs954895232 Chr.16: 81362661-81362661 on Build GRCh38
rs753746307 Chr.16: 81362488-81362488 on Build GRCh38
rs752087737 Chr.16: 81362647-81362648 on Build GRCh38
rs752661883 Chr.16: 81362546-81362546 on Build GRCh38
rs746599761 Chr.16: 81362471-81362471 on Build GRCh38
rs767206913 Chr.16: 81362658-81362658 on Build GRCh38
rs776907910 Chr.16: 81362608-81362608 on Build GRCh38
rs756800191 Chr.16: 81362640-81362640 on Build GRCh38
rs199908897 Chr.16: 81362518-81362518 on Build GRCh38
rs770708765 Chr.16: 81362473-81362473 on Build GRCh38
rs377288892 Chr.16: 81362470-81362470 on Build GRCh38
rs759342719 Chr.16: 81362540-81362540 on Build GRCh38
rs757918580 Chr.16: 81362466-81362466 on Build GRCh38
rs777349334 Chr.16: 81362553-81362553 on Build GRCh38
rs201451382 Chr.16: 81362542-81362542 on Build GRCh38
rs759128138 Chr.16: 81362632-81362632 on Build GRCh38
rs755366433 Chr.16: 81362576-81362576 on Build GRCh38
rs752616238 Chr.16: 81362465-81362465 on Build GRCh38
rs779203584 Chr.16: 81362580-81362580 on Build GRCh38
rs773225397 Chr.16: 81362591-81362591 on Build GRCh38
rs368407471 Chr.16: 81362573-81362573 on Build GRCh38
rs573350534 Chr.16: 81362669-81362669 on Build GRCh38
rs751149887 Chr.16: 81362563-81362563 on Build GRCh38
rs890618913 Chr.16: 81362579-81362579 on Build GRCh38
rs747068103 Chr.16: 81362594-81362594 on Build GRCh38
rs149373971 Chr.16: 81362490-81362490 on Build GRCh38
rs996280683 Chr.16: 81362486-81362486 on Build GRCh38
rs374004725 Chr.16: 81362570-81362570 on Build GRCh38
rs765020388 Chr.16: 81362615-81362615 on Build GRCh38
rs745551032 Chr.16: 81362575-81362575 on Build GRCh38
rs748632070 Chr.16: 81362587-81362587 on Build GRCh38
rs561109989 Chr.16: 81362631-81362634 on Build GRCh38
rs867264632 Chr.16: 81362551-81362551 on Build GRCh38
rs778888503 Chr.16: 81362461-81362461 on Build GRCh38
rs766772301 Chr.16: 81362619-81362619 on Build GRCh38
rs370426264 Chr.16: 81362590-81362590 on Build GRCh38
rs754108244 Chr.16: 81362529-81362529 on Build GRCh38
rs779272184 Chr.16: 81362663-81362663 on Build GRCh38
rs956021392 Chr.16: 81362592-81362592 on Build GRCh38
rs780528778 Chr.16: 81362487-81362487 on Build GRCh38
rs587779384 Chr.16: 81362609-81362609 on Build GRCh38
rs540478179 Chr.16: 81362678-81362678 on Build GRCh38
rs773294836 Chr.16: 81362501-81362501 on Build GRCh38
rs762904814 Chr.16: 81362620-81362620 on Build GRCh38
rs895658385 Chr.16: 81362512-81362512 on Build GRCh38
rs78861333 Chr.16: 81362662-81362662 on Build GRCh38
rs766607608 Chr.16: 81362522-81362522 on Build GRCh38
rs569189562 Chr.16: 81362558-81362558 on Build GRCh38
rs758279856 Chr.16: 81362552-81362552 on Build GRCh38

View Full Product Details