Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51980478-51980751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD14A
Entrez Gene ID
25864
Gene Name
abhydrolase domain containing 14A
Gene Aliases
DORZ1
UniGene
Hs.534400
Gene Chromosome Location
Chr.3: 51975026-51981200 on Build GRCh38
Transcripts
NM_015407.4 NM_001316331.1

Gene Symbol
ABHD14A-ACY1
Entrez Gene ID
100526760
Gene Name
ABHD14A-ACY1 readthrough
Gene Aliases
-
UniGene
Hs.334707
Gene Chromosome Location
Chr.3: 51975026-51989202 on Build GRCh38
Transcripts
NM_015407.4 NM_001316331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6861326 c.485T>C p.V162A Chr.3: 51980480-51980480 on Build GRCh38
2852423 c.535C>T p.P179S Chr.3: 51980530-51980530 on Build GRCh38
2852424 c.547C>T p.R183* Chr.3: 51980542-51980542 on Build GRCh38
4118933 c.626C>T p.A209V Chr.3: 51980621-51980621 on Build GRCh38
158821 c.534G>A p.L178L Chr.3: 51980529-51980529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144897211 Chr.3: 51980561-51980561 on Build GRCh38
rs749390049 Chr.3: 51980680-51980680 on Build GRCh38
rs906016189 Chr.3: 51980581-51980581 on Build GRCh38
rs752152340 Chr.3: 51980643-51980643 on Build GRCh38
rs752344825 Chr.3: 51980543-51980543 on Build GRCh38
rs1010338034 Chr.3: 51980692-51980692 on Build GRCh38
rs373822210 Chr.3: 51980661-51980661 on Build GRCh38
rs377621529 Chr.3: 51980677-51980677 on Build GRCh38
rs150338622 Chr.3: 51980505-51980505 on Build GRCh38
rs373076381 Chr.3: 51980629-51980629 on Build GRCh38
rs1018932278 Chr.3: 51980699-51980699 on Build GRCh38
rs375013972 Chr.3: 51980570-51980570 on Build GRCh38
rs370004391 Chr.3: 51980541-51980541 on Build GRCh38
rs764744391 Chr.3: 51980636-51980636 on Build GRCh38
rs761106676 Chr.3: 51980630-51980630 on Build GRCh38
rs918103477 Chr.3: 51980585-51980585 on Build GRCh38
rs764685951 Chr.3: 51980542-51980542 on Build GRCh38
rs762408906 Chr.3: 51980526-51980526 on Build GRCh38
rs1000403053 Chr.3: 51980613-51980613 on Build GRCh38
rs377611082 Chr.3: 51980667-51980667 on Build GRCh38
rs759424901 Chr.3: 51980628-51980628 on Build GRCh38
rs141581474 Chr.3: 51980578-51980578 on Build GRCh38
rs758000137 Chr.3: 51980545-51980545 on Build GRCh38
rs189631613 Chr.3: 51980551-51980551 on Build GRCh38
rs533685872 Chr.3: 51980722-51980722 on Build GRCh38
rs1012474109 Chr.3: 51980683-51980683 on Build GRCh38
rs926256502 Chr.3: 51980674-51980674 on Build GRCh38
rs566649968 Chr.3: 51980624-51980624 on Build GRCh38
rs768831036 Chr.3: 51980604-51980604 on Build GRCh38
rs141083259 Chr.3: 51980550-51980550 on Build GRCh38
rs753007139 Chr.3: 51980675-51980676 on Build GRCh38
rs774863477 Chr.3: 51980523-51980523 on Build GRCh38
rs139118990 Chr.3: 51980508-51980508 on Build GRCh38
rs763791077 Chr.3: 51980652-51980652 on Build GRCh38
rs762319081 Chr.3: 51980627-51980627 on Build GRCh38
rs781412372 Chr.3: 51980507-51980507 on Build GRCh38
rs749463097 Chr.3: 51980603-51980603 on Build GRCh38
rs753377764 Chr.3: 51980530-51980530 on Build GRCh38
rs756147060 Chr.3: 51980676-51980676 on Build GRCh38
rs768198518 Chr.3: 51980527-51980527 on Build GRCh38
rs371733964 Chr.3: 51980560-51980560 on Build GRCh38
rs3178064 Chr.3: 51980616-51980616 on Build GRCh38
rs763110053 Chr.3: 51980517-51980517 on Build GRCh38
rs376541544 Chr.3: 51980678-51980678 on Build GRCh38
rs769010826 Chr.3: 51980522-51980522 on Build GRCh38
rs767616415 Chr.3: 51980528-51980529 on Build GRCh38
rs369359084 Chr.3: 51980577-51980577 on Build GRCh38
rs775817685 Chr.3: 51980513-51980513 on Build GRCh38
rs762662695 Chr.3: 51980645-51980645 on Build GRCh38
rs780993829 Chr.3: 51980671-51980671 on Build GRCh38

View Full Product Details