Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15706330-15706587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF286A
Entrez Gene ID
57335
Gene Name
zinc finger protein 286A
Gene Aliases
ZNF286
UniGene
Hs.585799
Gene Chromosome Location
Chr.17: 15699577-15720787 on Build GRCh38
Transcripts
NM_001130842.1 NM_001288642.1 NM_001288643.1 NM_001288644.1 NM_001288645.1 NM_001288646.1 NM_001288647.1 NM_001288648.1 NM_001288649.1 NM_020652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146053 c.154A>C p.M52L Chr.17: 15706414-15706414 on Build GRCh38
3937260 c.187G>A p.D63N Chr.17: 15706447-15706447 on Build GRCh38
7423433 c.199A>T p.R67W Chr.17: 15706459-15706459 on Build GRCh38
975803 c.159C>T p.D53D Chr.17: 15706419-15706419 on Build GRCh38
5924700 c.167C>T p.P56L Chr.17: 15706427-15706427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763606346 Chr.17: 15706502-15706502 on Build GRCh38
rs768721867 Chr.17: 15706423-15706423 on Build GRCh38
rs554398671 Chr.17: 15706365-15706365 on Build GRCh38
rs149952794 Chr.17: 15706488-15706488 on Build GRCh38
rs201213141 Chr.17: 15706440-15706440 on Build GRCh38
rs768852871 Chr.17: 15706464-15706466 on Build GRCh38
rs755101350 Chr.17: 15706539-15706539 on Build GRCh38
rs1008058120 Chr.17: 15706378-15706378 on Build GRCh38
rs559208977 Chr.17: 15706399-15706399 on Build GRCh38
rs561098137 Chr.17: 15706560-15706560 on Build GRCh38
rs746180494 Chr.17: 15706414-15706414 on Build GRCh38
rs752382790 Chr.17: 15706390-15706390 on Build GRCh38
rs374168281 Chr.17: 15706510-15706510 on Build GRCh38
rs773079564 Chr.17: 15706455-15706455 on Build GRCh38
rs924996157 Chr.17: 15706561-15706561 on Build GRCh38
rs747166402 Chr.17: 15706378-15706378 on Build GRCh38
rs755822919 Chr.17: 15706500-15706500 on Build GRCh38
rs952125982 Chr.17: 15706450-15706450 on Build GRCh38
rs767112566 Chr.17: 15706479-15706479 on Build GRCh38
rs138466250 Chr.17: 15706370-15706370 on Build GRCh38
rs200603999 Chr.17: 15706537-15706537 on Build GRCh38
rs776900195 Chr.17: 15706436-15706436 on Build GRCh38
rs779306821 Chr.17: 15706401-15706401 on Build GRCh38
rs758967807 Chr.17: 15706416-15706416 on Build GRCh38
rs980901337 Chr.17: 15706452-15706452 on Build GRCh38
rs774426118 Chr.17: 15706470-15706470 on Build GRCh38
rs767343911 Chr.17: 15706388-15706388 on Build GRCh38
rs372037998 Chr.17: 15706456-15706456 on Build GRCh38
rs367996379 Chr.17: 15706443-15706443 on Build GRCh38
rs780607930 Chr.17: 15706513-15706513 on Build GRCh38
rs199895526 Chr.17: 15706418-15706418 on Build GRCh38
rs545764883 Chr.17: 15706377-15706377 on Build GRCh38
rs371458811 Chr.17: 15706478-15706478 on Build GRCh38
rs80140467 Chr.17: 15706454-15706454 on Build GRCh38
rs758695730 Chr.17: 15706511-15706511 on Build GRCh38
rs781221275 Chr.17: 15706540-15706540 on Build GRCh38
rs541431750 Chr.17: 15706541-15706541 on Build GRCh38

View Full Product Details