Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 13877489-13877714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEL1L2
Entrez Gene ID
80343
Gene Name
SEL1L2 ERAD E3 ligase adaptor subunit
Gene Aliases
C20orf50
UniGene
Hs.590879
Gene Chromosome Location
Chr.20: 13849188-13995333 on Build GRCh38
Transcripts
NM_001271539.1 NR_073206.1 NR_073207.1 XM_006723646.2 XM_006723647.2 XM_006723648.2 XM_006723650.2 XM_006723651.2 XM_006723652.2 XM_006723654.3 XM_006723655.2 XM_006723656.2 XM_011529374.1 XM_011529375.1 XM_011529376.1 XM_011529377.1 XM_011529378.1 XM_011529379.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
326436 c.1019T>C p.I340T Chr.20: 13877527-13877527 on Build GRCh38
3544181 c.975C>A p.F325L Chr.20: 13877571-13877571 on Build GRCh38
5391584 c.971A>G p.Y324C Chr.20: 13877575-13877575 on Build GRCh38
6774381 c.1001C>T p.A334V Chr.20: 13877545-13877545 on Build GRCh38
1318858 c.1022G>A p.G341E Chr.20: 13877524-13877524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029894969 Chr.20: 13877660-13877660 on Build GRCh38
rs771674811 Chr.20: 13877631-13877631 on Build GRCh38
rs940911188 Chr.20: 13877655-13877655 on Build GRCh38
rs773871457 Chr.20: 13877671-13877671 on Build GRCh38
rs781535936 Chr.20: 13877578-13877578 on Build GRCh38
rs770424703 Chr.20: 13877591-13877591 on Build GRCh38
rs749690152 Chr.20: 13877604-13877604 on Build GRCh38
rs773682692 Chr.20: 13877547-13877547 on Build GRCh38
rs200683153 Chr.20: 13877553-13877553 on Build GRCh38
rs770611148 Chr.20: 13877552-13877552 on Build GRCh38
rs985737149 Chr.20: 13877692-13877692 on Build GRCh38
rs751560376 Chr.20: 13877574-13877574 on Build GRCh38
rs778081202 Chr.20: 13877565-13877565 on Build GRCh38
rs768169027 Chr.20: 13877606-13877606 on Build GRCh38
rs762623161 Chr.20: 13877543-13877543 on Build GRCh38
rs199635097 Chr.20: 13877564-13877564 on Build GRCh38
rs895071166 Chr.20: 13877538-13877538 on Build GRCh38
rs761228877 Chr.20: 13877610-13877610 on Build GRCh38
rs746319532 Chr.20: 13877582-13877582 on Build GRCh38
rs958889283 Chr.20: 13877548-13877548 on Build GRCh38
rs768382921 Chr.20: 13877544-13877544 on Build GRCh38
rs373099694 Chr.20: 13877555-13877555 on Build GRCh38
rs780522526 Chr.20: 13877601-13877601 on Build GRCh38
rs772725721 Chr.20: 13877640-13877640 on Build GRCh38
rs776010648 Chr.20: 13877537-13877537 on Build GRCh38
rs766105501 Chr.20: 13877557-13877557 on Build GRCh38
rs909475260 Chr.20: 13877616-13877616 on Build GRCh38
rs761432871 Chr.20: 13877551-13877551 on Build GRCh38
rs138905672 Chr.20: 13877608-13877608 on Build GRCh38
rs574283400 Chr.20: 13877689-13877689 on Build GRCh38
rs753636072 Chr.20: 13877561-13877561 on Build GRCh38
rs762417122 Chr.20: 13877569-13877571 on Build GRCh38
rs757423633 Chr.20: 13877576-13877576 on Build GRCh38

View Full Product Details