Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30906491-30906740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEDAG
Entrez Gene ID
84935
Gene Name
mesenteric estrogen dependent adipogenesis
Gene Aliases
AWMS3, C13orf33, MEDA-4, MEDA4, hAWMS3
UniGene
Hs.147880
Gene Chromosome Location
Chr.13: 30906175-30925577 on Build GRCh38
Transcripts
NM_032849.3 NR_038287.1

Gene Symbol
TEX26-AS1
Entrez Gene ID
100507064
Gene Name
TEX26 antisense RNA 1
Gene Aliases
LINC00447
UniGene
Hs.654878
Gene Chromosome Location
Chr.13: 30882835-30932608 on Build GRCh38
Transcripts
NM_032849.3 NR_038287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6335731 c.69C>T p.R23R Chr.13: 30906584-30906584 on Build GRCh38
4608490 c.97G>T p.A33S Chr.13: 30906612-30906612 on Build GRCh38
6352610 c.174C>T p.A58A Chr.13: 30906689-30906689 on Build GRCh38
6520962 c.80C>A p.T27N Chr.13: 30906595-30906595 on Build GRCh38
7098446 c.24G>A p.P8P Chr.13: 30906539-30906539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773196246 Chr.13: 30906693-30906693 on Build GRCh38
rs762523878 Chr.13: 30906716-30906716 on Build GRCh38
rs200214346 Chr.13: 30906533-30906533 on Build GRCh38
rs1006607551 Chr.13: 30906695-30906695 on Build GRCh38
rs199670958 Chr.13: 30906588-30906588 on Build GRCh38
rs1056580047 Chr.13: 30906571-30906571 on Build GRCh38
rs778021784 Chr.13: 30906568-30906568 on Build GRCh38
rs757605584 Chr.13: 30906719-30906719 on Build GRCh38
rs776912049 Chr.13: 30906579-30906579 on Build GRCh38
rs778279697 Chr.13: 30906660-30906660 on Build GRCh38
rs759088171 Chr.13: 30906639-30906639 on Build GRCh38
rs767436094 Chr.13: 30906606-30906606 on Build GRCh38
rs904771005 Chr.13: 30906607-30906607 on Build GRCh38
rs753474255 Chr.13: 30906623-30906623 on Build GRCh38
rs761746695 Chr.13: 30906526-30906526 on Build GRCh38
rs781711508 Chr.13: 30906720-30906720 on Build GRCh38
rs758844060 Chr.13: 30906659-30906659 on Build GRCh38
rs776372402 Chr.13: 30906699-30906699 on Build GRCh38
rs757630646 Chr.13: 30906666-30906666 on Build GRCh38
rs749327526 Chr.13: 30906681-30906681 on Build GRCh38
rs760442815 Chr.13: 30906696-30906696 on Build GRCh38
rs761869378 Chr.13: 30906605-30906605 on Build GRCh38
rs760451601 Chr.13: 30906618-30906618 on Build GRCh38
rs753747875 Chr.13: 30906608-30906608 on Build GRCh38
rs754478440 Chr.13: 30906545-30906545 on Build GRCh38
rs745558596 Chr.13: 30906714-30906715 on Build GRCh38
rs750189432 Chr.13: 30906529-30906529 on Build GRCh38
rs9531945 Chr.13: 30906690-30906690 on Build GRCh38
rs964600358 Chr.13: 30906721-30906721 on Build GRCh38
rs764760167 Chr.13: 30906643-30906643 on Build GRCh38
rs71436423 Chr.13: 30906583-30906583 on Build GRCh38
rs774468841 Chr.13: 30906597-30906597 on Build GRCh38
rs1044282570 Chr.13: 30906628-30906628 on Build GRCh38
rs776591666 Chr.13: 30906519-30906519 on Build GRCh38
rs748624460 Chr.13: 30906549-30906549 on Build GRCh38
rs774419939 Chr.13: 30906524-30906524 on Build GRCh38
rs751939702 Chr.13: 30906663-30906663 on Build GRCh38
rs766011702 Chr.13: 30906620-30906620 on Build GRCh38
rs780299554 Chr.13: 30906676-30906676 on Build GRCh38
rs764571813 Chr.13: 30906717-30906717 on Build GRCh38
rs964921245 Chr.13: 30906520-30906520 on Build GRCh38
rs752208437 Chr.13: 30906709-30906709 on Build GRCh38
rs574746656 Chr.13: 30906557-30906557 on Build GRCh38
rs779452041 Chr.13: 30906548-30906548 on Build GRCh38
rs1019425876 Chr.13: 30906704-30906704 on Build GRCh38
rs767183130 Chr.13: 30906527-30906527 on Build GRCh38
rs753349582 Chr.13: 30906541-30906541 on Build GRCh38
rs752155046 Chr.13: 30906649-30906649 on Build GRCh38
rs954990902 Chr.13: 30906539-30906539 on Build GRCh38
rs1037808910 Chr.13: 30906712-30906712 on Build GRCh38
rs758899551 Chr.13: 30906553-30906553 on Build GRCh38
rs765998379 Chr.13: 30906534-30906534 on Build GRCh38
rs758738581 Chr.13: 30906715-30906715 on Build GRCh38
rs770099315 Chr.13: 30906674-30906674 on Build GRCh38
rs1047640126 Chr.13: 30906677-30906677 on Build GRCh38
rs764811845 Chr.13: 30906700-30906700 on Build GRCh38
rs759626056 Chr.13: 30906523-30906523 on Build GRCh38
rs771067374 Chr.13: 30906573-30906573 on Build GRCh38
rs200311232 Chr.13: 30906672-30906672 on Build GRCh38

View Full Product Details