Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65345623-65345884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPF2
Entrez Gene ID
5977
Gene Name
double PHD fingers 2
Gene Aliases
REQ, UBID4, ubi-d4
UniGene
Hs.13495
Gene Chromosome Location
Chr.11: 65333754-65352980 on Build GRCh38
Transcripts
NM_006268.4 XM_005274149.2 XM_017018101.1 XM_017018102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4756590 c.715G>A p.E239K Chr.11: 65345743-65345743 on Build GRCh38
3986353 c.699G>A p.L233L Chr.11: 65345727-65345727 on Build GRCh38
4910643 c.681C>G p.H227Q Chr.11: 65345709-65345709 on Build GRCh38
7088002 c.644G>T p.G215V Chr.11: 65345672-65345672 on Build GRCh38
1628163 c.685G>T p.A229S Chr.11: 65345713-65345713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199512449 Chr.11: 65345721-65345721 on Build GRCh38
rs768132085 Chr.11: 65345660-65345660 on Build GRCh38
rs945000211 Chr.11: 65345737-65345737 on Build GRCh38
rs755112635 Chr.11: 65345761-65345761 on Build GRCh38
rs766555584 Chr.11: 65345739-65345739 on Build GRCh38
rs764435076 Chr.11: 65345650-65345652 on Build GRCh38
rs191368000 Chr.11: 65345654-65345654 on Build GRCh38
rs1028964066 Chr.11: 65345659-65345659 on Build GRCh38
rs750866861 Chr.11: 65345700-65345700 on Build GRCh38
rs975312630 Chr.11: 65345787-65345787 on Build GRCh38
rs745799949 Chr.11: 65345834-65345834 on Build GRCh38
rs774054134 Chr.11: 65345671-65345671 on Build GRCh38
rs752984829 Chr.11: 65345772-65345772 on Build GRCh38
rs751922422 Chr.11: 65345835-65345835 on Build GRCh38
rs761247048 Chr.11: 65345742-65345742 on Build GRCh38
rs756098798 Chr.11: 65345840-65345840 on Build GRCh38
rs756980534 Chr.11: 65345778-65345778 on Build GRCh38
rs762578638 Chr.11: 65345658-65345658 on Build GRCh38
rs762038511 Chr.11: 65345685-65345685 on Build GRCh38
rs922128986 Chr.11: 65345796-65345796 on Build GRCh38
rs778963899 Chr.11: 65345763-65345763 on Build GRCh38
rs781079354 Chr.11: 65345813-65345813 on Build GRCh38
rs550928421 Chr.11: 65345697-65345697 on Build GRCh38

View Full Product Details