The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143805.1 | NP_001137277.1 | 2 - 2 | 808 | 71 |
NM_001143806.1 | NP_001137278.1 | 2 - 2 | 1023 | 71 | |
NM_001143807.1 | NP_001137279.1 | 2 - 2 | 909 | 71 | |
NM_001143808.1 | NP_001137280.1 | 2 - 2 | 745 | 71 | |
NM_001143809.1 | NP_001137281.1 | 2 - 2 | 755 | 71 | |
NM_001143810.1 | NP_001137282.1 | 3 - 3 | 1038 | 71 | |
NM_001143811.1 | NP_001137283.1 | 4 - 4 | 1155 | 71 | |
NM_001143812.1 | NP_001137284.1 | 2 - 2 | 897 | 71 | |
NM_001143813.1 | NP_001137285.1 | 2 - 2 | 979 | 71 | |
NM_001143814.1 | NP_001137286.1 | 3 - 3 | 1104 | 71 | |
NM_001143816.1 | NP_001137288.1 | 2 - 2 | 1502 | 71 | |
NM_001709.4 | NP_001700.2 | 2 - 2 | 997 | 71 | |
NM_170731.4 | NP_733927.1 | 2 - 2 | 1319 | 71 | |
NM_170732.4 | NP_733928.1 | 2 - 2 | 1106 | 71 | |
NM_170733.3 | NP_733929.1 | 2 - 2 | 1009 | 71 | |
NM_170734.3 | NP_733930.1 | 2 - 2 | 957 | 71 | |
NM_170735.5 | NP_733931.1 | 1 - 1 | 1736 | 71 | |
XM_011520280.2 | XP_011518582.1 | 2 - 2 | 1903 | 71 | |
GenBank mRNA | AF400438.1 | - | 1 - 1 | 669 | 71 |
AK289763.1 | - | 2 - 2 | 805 | 71 | |
AK289853.1 | - | 2 - 2 | 706 | 71 | |
AY054392.2 | - | 2 - 2 | 1010 | 71 | |
AY054393.2 | - | 2 - 2 | 752 | 71 | |
AY054394.2 | - | 2 - 2 | 967 | 71 | |
AY054395.2 | - | 2 - 2 | 1050 | 71 | |
AY054396.2 | - | 2 - 2 | 808 | 71 | |
AY054397.2 | - | 2 - 2 | 891 | 71 | |
AY054398.2 | - | 2 - 2 | 938 | 71 | |
AY054399.2 | - | 3 - 3 | 823 | 71 | |
AY054400.2 | - | 2 - 2 | 924 | 71 | |
AY054406.3 | - | 3 - 3 | 648 | 71 | |
AY656701.1 | - | 1 - 1 | 651 | 71 | |
BC029795.1 | - | 2 - 2 | 982 | 71 | |
EF674517.1 | - | 2 - 2 | 808 | 71 | |
EF674518.1 | - | 2 - 2 | 1023 | 71 | |
EF674519.1 | - | 2 - 2 | 1106 | 71 | |
EF674520.1 | - | 2 - 2 | 909 | 71 | |
EF674521.1 | - | 2 - 2 | 1009 | 71 | |
EF689009.1 | - | 1 - 1 | 1750 | 71 | |
EF689010.1 | - | 2 - 2 | 1516 | 71 | |
EF689011.1 | - | 2 - 2 | 745 | 71 | |
EF689012.1 | - | 2 - 2 | 755 | 71 | |
EF689013.1 | - | 2 - 2 | 897 | 71 | |
EF689014.1 | - | 2 - 2 | 979 | 71 | |
EF689015.1 | - | 2 - 2 | 997 | 71 | |
EF689016.1 | - | 3 - 3 | 1104 | 71 | |
EF689017.1 | - | 3 - 3 | 856 | 71 | |
EF689018.1 | - | 2 - 2 | 957 | 71 | |
EF689019.1 | - | 3 - 3 | 1038 | 71 | |
EF689020.1 | - | 4 - 4 | 1155 | 71 | |
EF689021.1 | - | 2 - 2 | 1319 | 71 | |
KC855559.1 | - | 1 - 1 | 187 | 71 | |
KP229412.1 | - | 1 - 1 | 651 | 71 | |
KU177943.1 | - | 1 - 1 | 651 | 71 | |
KU177944.1 | - | 2 - 2 | 534 | 71 | |
KU177945.1 | - | 2 - 2 | 507 | 71 | |
M61176.1 | - | 1 - 1 | 1213 | 71 | |
X60201.1 | - | 2 - 2 | 947 | 71 | |
X91251.1 | - | 2 - 2 | 935 | 71 |