Assay Design



Gene Transcripts

Gene Symbol
VHL
Entrez Gene ID
540957
Gene Name
von Hippel-Lindau tumor suppressor
Gene Aliases
-
Chromosome Location
Chr.22: 16714931 - 16719806 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110019.1 NP_001103489.1 1 - 2 260 103

View Full Product Details