Assay Design



Gene Transcripts

Gene Symbol
MT3
Entrez Gene ID
613320
Gene Name
metallothionein 3
Gene Aliases
-
Chromosome Location
Chr.18: 24051099 - 24052541 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113304.2 NP_001106775.1 1 - 2 91 80
GenBank mRNA BC126773.1 - 1 - 2 92 80

View Full Product Details