Assay Design
Gene Transcripts
- Gene Symbol
- SOST
- Entrez Gene ID
-
490948
- Gene Name
- sclerostin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 19538713 - 19544650 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_548071.6 |
XP_548071.2 |
- |
1217 |
139 |
View Full Product Details