Assay Design



Gene Transcripts

Gene Symbol
foxo
Entrez Gene ID
41709
Gene Name
forkhead box, sub-group O
Gene Aliases
3143, Afx, CG3143, DFOXO, DfoxO, Dfoxo, DmQ95V5, Dmel\CG3143, Dmel_CG3143, FKHR, FOXO, FOXO1, FoxO, Foxo, Foxo1, Q95V55, anon-WO0118547.374, cg3143, dFOXO, dFoxO, dFoxo, dfoxO, dfoxo, fd88A, foxO, foxo|dFOXO
Chromosome Location
Chr.3R: 14056945 - 14087978 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038197
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275628.1 NP_001262557.1 2 - 3 513 63
NM_001316497.1 NP_001303426.1 3 - 4 638 63
NM_142073.5 NP_650330.3 3 - 4 771 63
NM_206482.3 NP_996204.1 3 - 4 638 63
NM_206483.3 NP_996205.1 3 - 4 1029 63
GenBank mRNA AF426831.1 - 3 - 4 621 63
AY089542.1 - 3 - 4 1006 63

View Full Product Details