Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081769.1 | NP_001075238.1 | 19 - 20 | 2429 | 76 |
XM_023651656.1 | XP_023507424.1 | - | 2429 | 76 | |
XM_023651657.1 | XP_023507425.1 | - | 2815 | 76 | |
GenBank mRNA | AB039864.1 | - | 1 - 2 | 180 | 76 |
AJ493657.1 | - | 1 - 2 | 143 | 76 | |
AY027883.1 | - | 19 - 20 | 2429 | 76 |