Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67631345-67631887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX10
Entrez Gene ID
347853
Gene Name
T-box 10
Gene Aliases
TBX13, TBX7
UniGene
Hs.454480
Gene Chromosome Location
Chr.11: 67631303-67639560 on Build GRCh38
Transcripts
NM_005995.4 XM_011544992.2 XM_011544993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3980619 c.982C>G p.Q328E Chr.11: 67631781-67631781 on Build GRCh38
6320149 c.977C>A p.T326K Chr.11: 67631786-67631786 on Build GRCh38
6363013 c.1124T>C p.V375A Chr.11: 67631639-67631639 on Build GRCh38
6347328 c.1044C>A p.Y348* Chr.11: 67631719-67631719 on Build GRCh38
4912205 c.1121C>T p.T374I Chr.11: 67631642-67631642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761628835 Chr.11: 67631808-67631808 on Build GRCh38
rs775191487 Chr.11: 67631791-67631791 on Build GRCh38
rs766044725 Chr.11: 67631604-67631604 on Build GRCh38
rs199538876 Chr.11: 67631708-67631708 on Build GRCh38
rs912601042 Chr.11: 67631462-67631462 on Build GRCh38
rs531906539 Chr.11: 67631786-67631786 on Build GRCh38
rs945019177 Chr.11: 67631537-67631537 on Build GRCh38
rs368650048 Chr.11: 67631494-67631494 on Build GRCh38
rs767377721 Chr.11: 67631815-67631815 on Build GRCh38
rs755019737 Chr.11: 67631760-67631760 on Build GRCh38
rs768029522 Chr.11: 67631589-67631589 on Build GRCh38
rs572845935 Chr.11: 67631376-67631376 on Build GRCh38
rs754716841 Chr.11: 67631611-67631611 on Build GRCh38
rs764900930 Chr.11: 67631716-67631716 on Build GRCh38
rs951281990 Chr.11: 67631711-67631711 on Build GRCh38
rs929932357 Chr.11: 67631439-67631439 on Build GRCh38
rs757697215 Chr.11: 67631863-67631863 on Build GRCh38
rs369161057 Chr.11: 67631825-67631825 on Build GRCh38
rs778321213 Chr.11: 67631773-67631773 on Build GRCh38
rs780150971 Chr.11: 67631643-67631643 on Build GRCh38
rs774829938 Chr.11: 67631587-67631587 on Build GRCh38
rs750374429 Chr.11: 67631818-67631818 on Build GRCh38
rs560683911 Chr.11: 67631522-67631522 on Build GRCh38
rs758155529 Chr.11: 67631625-67631625 on Build GRCh38
rs962880679 Chr.11: 67631394-67631394 on Build GRCh38
rs267603142 Chr.11: 67631691-67631691 on Build GRCh38
rs962521492 Chr.11: 67631675-67631675 on Build GRCh38
rs763930638 Chr.11: 67631797-67631797 on Build GRCh38
rs772336874 Chr.11: 67631667-67631667 on Build GRCh38
rs1016503224 Chr.11: 67631620-67631620 on Build GRCh38
rs201389909 Chr.11: 67631729-67631729 on Build GRCh38
rs993461800 Chr.11: 67631646-67631646 on Build GRCh38
rs757238456 Chr.11: 67631749-67631749 on Build GRCh38
rs943262112 Chr.11: 67631503-67631503 on Build GRCh38
rs748108566 Chr.11: 67631443-67631444 on Build GRCh38
rs527776441 Chr.11: 67631639-67631639 on Build GRCh38
rs747335405 Chr.11: 67631860-67631860 on Build GRCh38
rs979831559 Chr.11: 67631383-67631383 on Build GRCh38
rs146672518 Chr.11: 67631702-67631702 on Build GRCh38
rs372134674 Chr.11: 67631569-67631569 on Build GRCh38
rs755072160 Chr.11: 67631820-67631820 on Build GRCh38
rs752485816 Chr.11: 67631616-67631616 on Build GRCh38
rs1531514 Chr.11: 67631391-67631391 on Build GRCh38
rs143461859 Chr.11: 67631471-67631471 on Build GRCh38
rs777058708 Chr.11: 67631415-67631415 on Build GRCh38
rs758596700 Chr.11: 67631831-67631831 on Build GRCh38
rs4024255 Chr.11: 67631410-67631410 on Build GRCh38
rs751559619 Chr.11: 67631640-67631640 on Build GRCh38
rs775680202 Chr.11: 67631559-67631559 on Build GRCh38
rs915125877 Chr.11: 67631734-67631734 on Build GRCh38
rs370073114 Chr.11: 67631719-67631719 on Build GRCh38
rs748713696 Chr.11: 67631544-67631544 on Build GRCh38
rs541836460 Chr.11: 67631514-67631514 on Build GRCh38
rs1012945434 Chr.11: 67631365-67631365 on Build GRCh38
rs986713272 Chr.11: 67631453-67631453 on Build GRCh38
rs748377405 Chr.11: 67631767-67631767 on Build GRCh38
rs764916916 Chr.11: 67631612-67631612 on Build GRCh38
rs201107050 Chr.11: 67631806-67631806 on Build GRCh38
rs374679098 Chr.11: 67631849-67631849 on Build GRCh38
rs773597039 Chr.11: 67631591-67631591 on Build GRCh38
rs748480424 Chr.11: 67631662-67631662 on Build GRCh38
rs768981256 Chr.11: 67631573-67631573 on Build GRCh38
rs753876539 Chr.11: 67631759-67631759 on Build GRCh38
rs762108181 Chr.11: 67631588-67631588 on Build GRCh38
rs577211836 Chr.11: 67631443-67631443 on Build GRCh38
rs938902213 Chr.11: 67631571-67631571 on Build GRCh38
rs781132994 Chr.11: 67631755-67631755 on Build GRCh38
rs868037507 Chr.11: 67631861-67631861 on Build GRCh38
rs762746844 Chr.11: 67631733-67631733 on Build GRCh38
rs759893702 Chr.11: 67631586-67631586 on Build GRCh38
rs372881982 Chr.11: 67631828-67631828 on Build GRCh38
rs768875858 Chr.11: 67631658-67631658 on Build GRCh38
rs771514015 Chr.11: 67631779-67631779 on Build GRCh38
rs779037196 Chr.11: 67631661-67631661 on Build GRCh38
rs770532769 Chr.11: 67631703-67631703 on Build GRCh38
rs763866445 Chr.11: 67631737-67631737 on Build GRCh38
rs912442180 Chr.11: 67631756-67631756 on Build GRCh38
rs753535154 Chr.11: 67631607-67631607 on Build GRCh38
rs747494980 Chr.11: 67631777-67631777 on Build GRCh38
rs756186322 Chr.11: 67631642-67631642 on Build GRCh38
rs142856471 Chr.11: 67631785-67631785 on Build GRCh38
rs773756575 Chr.11: 67631681-67631681 on Build GRCh38
rs963345723 Chr.11: 67631378-67631378 on Build GRCh38
rs371576719 Chr.11: 67631505-67631505 on Build GRCh38
rs777696017 Chr.11: 67631636-67631636 on Build GRCh38
rs762557472 Chr.11: 67631795-67631795 on Build GRCh38
rs751471550 Chr.11: 67631738-67631738 on Build GRCh38
rs376486770 Chr.11: 67631580-67631580 on Build GRCh38
rs779139266 Chr.11: 67631761-67631761 on Build GRCh38
rs1026361656 Chr.11: 67631395-67631395 on Build GRCh38
rs749493214 Chr.11: 67631644-67631644 on Build GRCh38
rs772479926 Chr.11: 67631771-67631771 on Build GRCh38

View Full Product Details