Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68032201-68032705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFS8
Entrez Gene ID
4728
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
Gene Aliases
CI-23k, CI23KD, TYKY
UniGene
Hs.90443
Gene Chromosome Location
Chr.11: 68030617-68036647 on Build GRCh38
Transcripts
NM_002496.3 XM_005274013.1 XM_005274014.2 XM_011545053.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687592 c.58+2T>G p.? Chr.11: 68032211-68032211 on Build GRCh38
4036063 c.63T>C p.P21P Chr.11: 68032290-68032290 on Build GRCh38
3931419 c.103A>C p.T35P Chr.11: 68032330-68032330 on Build GRCh38
6723829 c.74G>A p.R25Q Chr.11: 68032301-68032301 on Build GRCh38
1605124 c.76A>G p.S26G Chr.11: 68032303-68032303 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs551690595 Chr.11: 68032501-68032501 on Build GRCh38
rs533915040 Chr.11: 68032270-68032270 on Build GRCh38
rs779201630 Chr.11: 68032305-68032305 on Build GRCh38
rs745923537 Chr.11: 68032351-68032351 on Build GRCh38
rs748087950 Chr.11: 68032312-68032312 on Build GRCh38
rs865986879 Chr.11: 68032292-68032292 on Build GRCh38
rs1019213622 Chr.11: 68032502-68032502 on Build GRCh38
rs139818945 Chr.11: 68032324-68032324 on Build GRCh38
rs549490599 Chr.11: 68032600-68032600 on Build GRCh38
rs533345614 Chr.11: 68032234-68032234 on Build GRCh38
rs996938610 Chr.11: 68032511-68032511 on Build GRCh38
rs372874720 Chr.11: 68032432-68032432 on Build GRCh38
rs750729830 Chr.11: 68032677-68032677 on Build GRCh38
rs960506638 Chr.11: 68032367-68032367 on Build GRCh38
rs767967357 Chr.11: 68032374-68032374 on Build GRCh38
rs889523221 Chr.11: 68032565-68032565 on Build GRCh38
rs760208390 Chr.11: 68032395-68032395 on Build GRCh38
rs747127279 Chr.11: 68032486-68032486 on Build GRCh38
rs537149486 Chr.11: 68032590-68032590 on Build GRCh38
rs748538081 Chr.11: 68032447-68032447 on Build GRCh38
rs946450900 Chr.11: 68032570-68032570 on Build GRCh38
rs563320012 Chr.11: 68032519-68032519 on Build GRCh38
rs1049532144 Chr.11: 68032534-68032534 on Build GRCh38
rs981214091 Chr.11: 68032632-68032632 on Build GRCh38
rs2075626 Chr.11: 68032459-68032459 on Build GRCh38
rs531754129 Chr.11: 68032434-68032434 on Build GRCh38
rs267603145 Chr.11: 68032306-68032306 on Build GRCh38
rs962014208 Chr.11: 68032664-68032664 on Build GRCh38
rs778922119 Chr.11: 68032311-68032311 on Build GRCh38
rs561492258 Chr.11: 68032300-68032300 on Build GRCh38
rs1044974378 Chr.11: 68032381-68032381 on Build GRCh38
rs1031177280 Chr.11: 68032577-68032577 on Build GRCh38
rs1023759513 Chr.11: 68032628-68032628 on Build GRCh38
rs898446425 Chr.11: 68032398-68032398 on Build GRCh38
rs976301824 Chr.11: 68032520-68032520 on Build GRCh38
rs769788621 Chr.11: 68032352-68032352 on Build GRCh38
rs775748305 Chr.11: 68032353-68032353 on Build GRCh38
rs546989001 Chr.11: 68032669-68032669 on Build GRCh38
rs940930199 Chr.11: 68032499-68032499 on Build GRCh38
rs776909632 Chr.11: 68032243-68032243 on Build GRCh38
rs150584898 Chr.11: 68032269-68032269 on Build GRCh38
rs71459685 Chr.11: 68032410-68032410 on Build GRCh38
rs573290810 Chr.11: 68032373-68032373 on Build GRCh38
rs368420184 Chr.11: 68032241-68032241 on Build GRCh38
rs753486707 Chr.11: 68032302-68032302 on Build GRCh38
rs576172614 Chr.11: 68032489-68032489 on Build GRCh38
rs774639417 Chr.11: 68032359-68032359 on Build GRCh38
rs1015351759 Chr.11: 68032651-68032651 on Build GRCh38
rs530783124 Chr.11: 68032594-68032594 on Build GRCh38
rs749069132 Chr.11: 68032225-68032225 on Build GRCh38
rs759701890 Chr.11: 68032245-68032245 on Build GRCh38
rs1046859167 Chr.11: 68032683-68032683 on Build GRCh38
rs932118304 Chr.11: 68032530-68032530 on Build GRCh38
rs769026952 Chr.11: 68032273-68032275 on Build GRCh38
rs777910193 Chr.11: 68032482-68032482 on Build GRCh38
rs968224404 Chr.11: 68032265-68032265 on Build GRCh38
rs918034213 Chr.11: 68032602-68032602 on Build GRCh38
rs369602258 Chr.11: 68032291-68032291 on Build GRCh38
rs1012026882 Chr.11: 68032625-68032625 on Build GRCh38
rs1010972536 Chr.11: 68032260-68032260 on Build GRCh38
rs948077907 Chr.11: 68032613-68032613 on Build GRCh38
rs371837015 Chr.11: 68032383-68032383 on Build GRCh38
rs752057064 Chr.11: 68032276-68032276 on Build GRCh38
rs1043794478 Chr.11: 68032574-68032574 on Build GRCh38
rs931226394 Chr.11: 68032649-68032649 on Build GRCh38
rs551598944 Chr.11: 68032235-68032235 on Build GRCh38
rs768355088 Chr.11: 68032226-68032226 on Build GRCh38
rs147455935 Chr.11: 68032301-68032301 on Build GRCh38
rs769051506 Chr.11: 68032356-68032356 on Build GRCh38
rs1046889854 Chr.11: 68032446-68032446 on Build GRCh38
rs769994523 Chr.11: 68032248-68032248 on Build GRCh38
rs115295367 Chr.11: 68032639-68032639 on Build GRCh38
rs749864835 Chr.11: 68032293-68032293 on Build GRCh38
rs140326349 Chr.11: 68032637-68032637 on Build GRCh38
rs952229821 Chr.11: 68032635-68032636 on Build GRCh38
rs769767821 Chr.11: 68032650-68032650 on Build GRCh38
rs369624943 Chr.11: 68032662-68032662 on Build GRCh38
rs968738236 Chr.11: 68032521-68032521 on Build GRCh38
rs979588634 Chr.11: 68032573-68032573 on Build GRCh38
rs963670423 Chr.11: 68032503-68032503 on Build GRCh38
rs776752329 Chr.11: 68032580-68032580 on Build GRCh38
rs3133266 Chr.11: 68032264-68032264 on Build GRCh38
rs777858020 Chr.11: 68032328-68032328 on Build GRCh38

View Full Product Details