Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68736452-68736956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TESMIN
Entrez Gene ID
9633
Gene Name
testis expressed metallothionein like protein
Gene Aliases
CXCDC2, MTL5, MTLT
UniGene
Hs.145932
Gene Chromosome Location
Chr.11: 68706674-68751564 on Build GRCh38
Transcripts
NM_001039656.1 NM_004923.3 XM_011545402.1 XM_011545403.2 XM_017018588.1

SNP Detail

SNP ID Chromosome Location
rs180920192 Chr.11: 68736902-68736902 on Build GRCh38
rs917405589 Chr.11: 68736717-68736717 on Build GRCh38
rs934208593 Chr.11: 68736893-68736893 on Build GRCh38
rs753934638 Chr.11: 68736662-68736662 on Build GRCh38
rs937434326 Chr.11: 68736653-68736653 on Build GRCh38
rs909511646 Chr.11: 68736537-68736537 on Build GRCh38
rs972954479 Chr.11: 68736544-68736544 on Build GRCh38
rs571115523 Chr.11: 68736711-68736712 on Build GRCh38
rs777250087 Chr.11: 68736710-68736711 on Build GRCh38
rs750022662 Chr.11: 68736636-68736637 on Build GRCh38
rs961018604 Chr.11: 68736809-68736809 on Build GRCh38
rs902504581 Chr.11: 68736692-68736692 on Build GRCh38
rs985594444 Chr.11: 68736896-68736896 on Build GRCh38
rs1022764689 Chr.11: 68736766-68736768 on Build GRCh38
rs978695046 Chr.11: 68736603-68736603 on Build GRCh38
rs968573434 Chr.11: 68736926-68736926 on Build GRCh38
rs750771675 Chr.11: 68736891-68736891 on Build GRCh38
rs1036633527 Chr.11: 68736804-68736804 on Build GRCh38
rs889875252 Chr.11: 68736606-68736606 on Build GRCh38
rs867935348 Chr.11: 68736610-68736610 on Build GRCh38
rs1014110260 Chr.11: 68736757-68736759 on Build GRCh38
rs1019892592 Chr.11: 68736909-68736909 on Build GRCh38
rs930973495 Chr.11: 68736853-68736853 on Build GRCh38
rs922876434 Chr.11: 68736847-68736847 on Build GRCh38
rs972984143 Chr.11: 68736814-68736814 on Build GRCh38
rs865786912 Chr.11: 68736543-68736543 on Build GRCh38
rs1043804729 Chr.11: 68736934-68736934 on Build GRCh38
rs761421045 Chr.11: 68736795-68736795 on Build GRCh38
rs764222222 Chr.11: 68736601-68736601 on Build GRCh38
rs922804493 Chr.11: 68736562-68736562 on Build GRCh38
rs993445014 Chr.11: 68736665-68736665 on Build GRCh38
rs970214708 Chr.11: 68736802-68736802 on Build GRCh38
rs934144591 Chr.11: 68736599-68736599 on Build GRCh38
rs984200577 Chr.11: 68736480-68736480 on Build GRCh38
rs1048049900 Chr.11: 68736890-68736890 on Build GRCh38
rs184985610 Chr.11: 68736918-68736918 on Build GRCh38
rs532308279 Chr.11: 68736843-68736843 on Build GRCh38
rs895711596 Chr.11: 68736738-68736738 on Build GRCh38
rs762066762 Chr.11: 68736719-68736719 on Build GRCh38
rs796345255 Chr.11: 68736480-68736482 on Build GRCh38
rs543923146 Chr.11: 68736925-68736925 on Build GRCh38
rs758833232 Chr.11: 68736911-68736911 on Build GRCh38

View Full Product Details