Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69703496-69703996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF19
Entrez Gene ID
9965
Gene Name
fibroblast growth factor 19
Gene Aliases
-
UniGene
Hs.249200
Gene Chromosome Location
Chr.11: 69698238-69704338 on Build GRCh38
Transcripts
NM_005117.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425571 c.95C>T p.P32L Chr.11: 69703782-69703782 on Build GRCh38
6552349 c.220C>A p.Q74K Chr.11: 69703657-69703657 on Build GRCh38
3935612 c.33G>T p.W11C Chr.11: 69703844-69703844 on Build GRCh38
6960243 c.160G>T p.G54W Chr.11: 69703717-69703717 on Build GRCh38
6968363 c.163C>T p.L55F Chr.11: 69703714-69703714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037647582 Chr.11: 69703775-69703775 on Build GRCh38
rs987180972 Chr.11: 69703938-69703938 on Build GRCh38
rs1013693090 Chr.11: 69703585-69703585 on Build GRCh38
rs987211751 Chr.11: 69703678-69703678 on Build GRCh38
rs555015996 Chr.11: 69703598-69703598 on Build GRCh38
rs1040281286 Chr.11: 69703740-69703740 on Build GRCh38
rs999316374 Chr.11: 69703857-69703857 on Build GRCh38
rs573160787 Chr.11: 69703600-69703600 on Build GRCh38
rs528044141 Chr.11: 69703918-69703918 on Build GRCh38
rs1024792578 Chr.11: 69703589-69703589 on Build GRCh38
rs988825791 Chr.11: 69703955-69703955 on Build GRCh38
rs1046522275 Chr.11: 69703555-69703555 on Build GRCh38
rs1002113100 Chr.11: 69703607-69703607 on Build GRCh38
rs563123516 Chr.11: 69703821-69703821 on Build GRCh38
rs540968622 Chr.11: 69703606-69703606 on Build GRCh38
rs905024850 Chr.11: 69703841-69703841 on Build GRCh38
rs1030129842 Chr.11: 69703847-69703847 on Build GRCh38
rs868553800 Chr.11: 69703586-69703586 on Build GRCh38
rs569437357 Chr.11: 69703959-69703959 on Build GRCh38
rs557621967 Chr.11: 69703954-69703954 on Build GRCh38
rs939925445 Chr.11: 69703599-69703599 on Build GRCh38
rs928104252 Chr.11: 69703967-69703967 on Build GRCh38
rs1031163552 Chr.11: 69703861-69703861 on Build GRCh38
rs376666157 Chr.11: 69703851-69703851 on Build GRCh38
rs571953909 Chr.11: 69703952-69703952 on Build GRCh38
rs948250492 Chr.11: 69703539-69703539 on Build GRCh38
rs892484058 Chr.11: 69703787-69703787 on Build GRCh38
rs753330387 Chr.11: 69703907-69703907 on Build GRCh38
rs928072803 Chr.11: 69703647-69703647 on Build GRCh38
rs969236533 Chr.11: 69703596-69703596 on Build GRCh38
rs114327249 Chr.11: 69703819-69703819 on Build GRCh38
rs1046435798 Chr.11: 69703966-69703966 on Build GRCh38
rs548634466 Chr.11: 69703854-69703854 on Build GRCh38
rs919766948 Chr.11: 69703613-69703613 on Build GRCh38
rs972938511 Chr.11: 69703604-69703604 on Build GRCh38
rs888488526 Chr.11: 69703700-69703700 on Build GRCh38
rs899059742 Chr.11: 69703908-69703908 on Build GRCh38
rs1052422715 Chr.11: 69703781-69703781 on Build GRCh38
rs1047515735 Chr.11: 69703665-69703665 on Build GRCh38
rs559561285 Chr.11: 69703774-69703774 on Build GRCh38
rs954444372 Chr.11: 69703663-69703663 on Build GRCh38
rs530553980 Chr.11: 69703825-69703825 on Build GRCh38
rs889553161 Chr.11: 69703628-69703628 on Build GRCh38
rs1001998520 Chr.11: 69703873-69703873 on Build GRCh38
rs1049476287 Chr.11: 69703622-69703622 on Build GRCh38
rs566945997 Chr.11: 69703896-69703896 on Build GRCh38
rs925526460 Chr.11: 69703767-69703767 on Build GRCh38
rs1017270999 Chr.11: 69703941-69703941 on Build GRCh38
rs892520788 Chr.11: 69703561-69703561 on Build GRCh38
rs538941127 Chr.11: 69703953-69703953 on Build GRCh38
rs931172575 Chr.11: 69703618-69703618 on Build GRCh38
rs546268599 Chr.11: 69703949-69703949 on Build GRCh38
rs1016220726 Chr.11: 69703633-69703633 on Build GRCh38
rs912723093 Chr.11: 69703681-69703681 on Build GRCh38
rs543125202 Chr.11: 69703559-69703559 on Build GRCh38
rs768424122 Chr.11: 69703965-69703965 on Build GRCh38
rs533068764 Chr.11: 69703814-69703814 on Build GRCh38
rs993846387 Chr.11: 69703912-69703912 on Build GRCh38
rs767954150 Chr.11: 69703760-69703760 on Build GRCh38
rs963626963 Chr.11: 69703634-69703634 on Build GRCh38
rs930040226 Chr.11: 69703649-69703649 on Build GRCh38
rs942913626 Chr.11: 69703715-69703715 on Build GRCh38
rs954412146 Chr.11: 69703927-69703927 on Build GRCh38
rs1005963395 Chr.11: 69703717-69703717 on Build GRCh38
rs751688902 Chr.11: 69703516-69703516 on Build GRCh38

View Full Product Details