Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74696180-74696678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
UniGene
Hs.432379
Gene Chromosome Location
Chr.11: 74696428-74731385 on Build GRCh38
Transcripts
NM_001278473.2 NM_001304390.1 NM_001304391.1 NM_001304415.1 NM_001304416.1 NM_001304417.1 NM_015424.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3980671 c.1301G>C p.G434A Chr.11: 74696553-74696553 on Build GRCh38
5765457 c.1325G>A p.R442K Chr.11: 74696529-74696529 on Build GRCh38
3870203 c.1301G>A p.G434E Chr.11: 74696553-74696553 on Build GRCh38
1628220 c.1340A>G p.D447G Chr.11: 74696514-74696514 on Build GRCh38
7286019 c.1286C>G p.P429R Chr.11: 74696568-74696568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569575109 Chr.11: 74696551-74696551 on Build GRCh38
rs369700929 Chr.11: 74696632-74696632 on Build GRCh38
rs765337940 Chr.11: 74696588-74696588 on Build GRCh38
rs779673508 Chr.11: 74696549-74696549 on Build GRCh38
rs921348050 Chr.11: 74696628-74696628 on Build GRCh38
rs765248093 Chr.11: 74696508-74696508 on Build GRCh38
rs557088622 Chr.11: 74696257-74696257 on Build GRCh38
rs753597569 Chr.11: 74696526-74696529 on Build GRCh38
rs768700793 Chr.11: 74696559-74696559 on Build GRCh38
rs761517458 Chr.11: 74696573-74696573 on Build GRCh38
rs142704636 Chr.11: 74696520-74696520 on Build GRCh38
rs758401171 Chr.11: 74696214-74696214 on Build GRCh38
rs771104143 Chr.11: 74696389-74696389 on Build GRCh38
rs368427060 Chr.11: 74696215-74696215 on Build GRCh38
rs867076152 Chr.11: 74696249-74696249 on Build GRCh38
rs1032777985 Chr.11: 74696440-74696440 on Build GRCh38
rs191985158 Chr.11: 74696541-74696541 on Build GRCh38
rs4539352 Chr.11: 74696224-74696224 on Build GRCh38
rs549440756 Chr.11: 74696239-74696239 on Build GRCh38
rs755799240 Chr.11: 74696630-74696630 on Build GRCh38
rs749279858 Chr.11: 74696554-74696554 on Build GRCh38
rs986762076 Chr.11: 74696658-74696658 on Build GRCh38
rs759761087 Chr.11: 74696587-74696587 on Build GRCh38
rs774352781 Chr.11: 74696569-74696569 on Build GRCh38
rs3819242 Chr.11: 74696631-74696631 on Build GRCh38
rs149731074 Chr.11: 74696574-74696574 on Build GRCh38
rs762836827 Chr.11: 74696578-74696578 on Build GRCh38
rs774264579 Chr.11: 74696459-74696459 on Build GRCh38
rs757576780 Chr.11: 74696617-74696617 on Build GRCh38
rs184393291 Chr.11: 74696618-74696618 on Build GRCh38
rs749631194 Chr.11: 74696309-74696309 on Build GRCh38
rs980598765 Chr.11: 74696533-74696533 on Build GRCh38
rs769211166 Chr.11: 74696548-74696548 on Build GRCh38
rs571865749 Chr.11: 74696534-74696534 on Build GRCh38
rs776736799 Chr.11: 74696575-74696575 on Build GRCh38
rs759545166 Chr.11: 74696482-74696482 on Build GRCh38
rs774574018 Chr.11: 74696488-74696488 on Build GRCh38
rs140967711 Chr.11: 74696256-74696256 on Build GRCh38
rs766505635 Chr.11: 74696467-74696467 on Build GRCh38
rs761486314 Chr.11: 74696518-74696518 on Build GRCh38
rs974534752 Chr.11: 74696625-74696625 on Build GRCh38
rs886888739 Chr.11: 74696589-74696589 on Build GRCh38
rs763513527 Chr.11: 74696609-74696609 on Build GRCh38
rs1012575186 Chr.11: 74696531-74696531 on Build GRCh38
rs753232281 Chr.11: 74696512-74696512 on Build GRCh38
rs552005307 Chr.11: 74696343-74696343 on Build GRCh38
rs745469119 Chr.11: 74696546-74696546 on Build GRCh38
rs939080198 Chr.11: 74696553-74696553 on Build GRCh38
rs1008810137 Chr.11: 74696242-74696242 on Build GRCh38
rs535457875 Chr.11: 74696616-74696616 on Build GRCh38
rs751859814 Chr.11: 74696537-74696537 on Build GRCh38
rs754024812 Chr.11: 74696469-74696469 on Build GRCh38
rs768604730 Chr.11: 74696455-74696455 on Build GRCh38
rs374354944 Chr.11: 74696610-74696610 on Build GRCh38
rs139734110 Chr.11: 74696513-74696513 on Build GRCh38
rs140831310 Chr.11: 74696540-74696540 on Build GRCh38
rs956677231 Chr.11: 74696345-74696345 on Build GRCh38
rs187489005 Chr.11: 74696644-74696644 on Build GRCh38
rs941026936 Chr.11: 74696602-74696602 on Build GRCh38
rs776028855 Chr.11: 74696488-74696489 on Build GRCh38
rs115692087 Chr.11: 74696539-74696539 on Build GRCh38
rs989910364 Chr.11: 74696348-74696348 on Build GRCh38
rs528755102 Chr.11: 74696302-74696302 on Build GRCh38
rs958585136 Chr.11: 74696466-74696468 on Build GRCh38
rs773056228 Chr.11: 74696575-74696584 on Build GRCh38
rs529272339 Chr.11: 74696236-74696236 on Build GRCh38
rs752754207 Chr.11: 74696608-74696608 on Build GRCh38
rs761013868 Chr.11: 74696461-74696461 on Build GRCh38

View Full Product Details