Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75271562-75272089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRB1
Entrez Gene ID
408
Gene Name
arrestin beta 1
Gene Aliases
ARB1, ARR1
UniGene
Hs.503284
Gene Chromosome Location
Chr.11: 75260122-75351831 on Build GRCh38
Transcripts
NM_004041.4 NM_020251.3 XM_011545034.2 XM_011545035.2 XM_017017750.1 XM_017017751.1 XM_017017752.1 XM_017017753.1 XM_017017754.1

SNP Detail

SNP ID Chromosome Location
rs976606700 Chr.11: 75271744-75271744 on Build GRCh38
rs905990863 Chr.11: 75271836-75271836 on Build GRCh38
rs1037658595 Chr.11: 75271765-75271765 on Build GRCh38
rs985189559 Chr.11: 75271918-75271918 on Build GRCh38
rs34439199 Chr.11: 75271943-75271943 on Build GRCh38
rs1044464305 Chr.11: 75272056-75272056 on Build GRCh38
rs147166849 Chr.11: 75272026-75272026 on Build GRCh38
rs530971363 Chr.11: 75271846-75271846 on Build GRCh38
rs776392603 Chr.11: 75271671-75271671 on Build GRCh38
rs35666856 Chr.11: 75271750-75271750 on Build GRCh38
rs766142423 Chr.11: 75271671-75271671 on Build GRCh38
rs2276309 Chr.11: 75271880-75271880 on Build GRCh38
rs1037224455 Chr.11: 75272025-75272025 on Build GRCh38
rs759410112 Chr.11: 75271917-75271917 on Build GRCh38
rs35895385 Chr.11: 75271985-75271985 on Build GRCh38
rs1056847726 Chr.11: 75271821-75271821 on Build GRCh38
rs909980322 Chr.11: 75271990-75271990 on Build GRCh38
rs922673631 Chr.11: 75271647-75271647 on Build GRCh38
rs185595523 Chr.11: 75272059-75272059 on Build GRCh38
rs188909651 Chr.11: 75271910-75271910 on Build GRCh38
rs778726709 Chr.11: 75271692-75271692 on Build GRCh38
rs941500055 Chr.11: 75272022-75272022 on Build GRCh38
rs539036864 Chr.11: 75271859-75271859 on Build GRCh38
rs867574038 Chr.11: 75271725-75271725 on Build GRCh38
rs956207739 Chr.11: 75271613-75271613 on Build GRCh38
rs36123903 Chr.11: 75271838-75271838 on Build GRCh38
rs893239978 Chr.11: 75271815-75271816 on Build GRCh38
rs867318415 Chr.11: 75271718-75271718 on Build GRCh38
rs36004313 Chr.11: 75271878-75271878 on Build GRCh38
rs2276310 Chr.11: 75271895-75271895 on Build GRCh38
rs572257591 Chr.11: 75271907-75271907 on Build GRCh38
rs754840432 Chr.11: 75271687-75271687 on Build GRCh38
rs772912852 Chr.11: 75271817-75271817 on Build GRCh38
rs137925861 Chr.11: 75271686-75271686 on Build GRCh38
rs993551041 Chr.11: 75271820-75271820 on Build GRCh38
rs35891965 Chr.11: 75272038-75272038 on Build GRCh38
rs748170131 Chr.11: 75271703-75271703 on Build GRCh38
rs965219900 Chr.11: 75271969-75271969 on Build GRCh38
rs777381801 Chr.11: 75271724-75271724 on Build GRCh38
rs181649297 Chr.11: 75272018-75272018 on Build GRCh38
rs956007709 Chr.11: 75271896-75271896 on Build GRCh38
rs866207683 Chr.11: 75271701-75271701 on Build GRCh38
rs1053109114 Chr.11: 75271778-75271778 on Build GRCh38
rs985723282 Chr.11: 75271602-75271602 on Build GRCh38
rs1022724420 Chr.11: 75271858-75271858 on Build GRCh38
rs371736924 Chr.11: 75271764-75271764 on Build GRCh38
rs555537326 Chr.11: 75271902-75271902 on Build GRCh38
rs372256796 Chr.11: 75272055-75272055 on Build GRCh38
rs976554189 Chr.11: 75271986-75271986 on Build GRCh38
rs935250579 Chr.11: 75272023-75272023 on Build GRCh38
rs757838584 Chr.11: 75271720-75271720 on Build GRCh38
rs770729011 Chr.11: 75271762-75271762 on Build GRCh38
rs976799830 Chr.11: 75271632-75271632 on Build GRCh38
rs768279064 Chr.11: 75271652-75271652 on Build GRCh38
rs954045827 Chr.11: 75271933-75271933 on Build GRCh38

View Full Product Details