Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86464964-86465471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ME3
Entrez Gene ID
10873
Gene Name
malic enzyme 3
Gene Aliases
NADP-ME
UniGene
Hs.199743
Gene Chromosome Location
Chr.11: 86441108-86674745 on Build GRCh38
Transcripts
NM_001014811.1 NM_001161586.1 NM_006680.2 XM_005273717.1 XM_017017136.1 XM_017017137.1 XM_017017138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641581 c.866G>A p.R289H Chr.11: 86465144-86465144 on Build GRCh38
932737 c.861C>A p.A287A Chr.11: 86465149-86465149 on Build GRCh38
932738 c.820A>C p.N274H Chr.11: 86465190-86465190 on Build GRCh38
6710468 c.894C>T p.Y298Y Chr.11: 86465116-86465116 on Build GRCh38
5721766 c.882C>T p.Y294Y Chr.11: 86465128-86465128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752542803 Chr.11: 86465136-86465136 on Build GRCh38
rs543008074 Chr.11: 86465398-86465398 on Build GRCh38
rs112485429 Chr.11: 86465388-86465388 on Build GRCh38
rs1016071276 Chr.11: 86465416-86465416 on Build GRCh38
rs560527957 Chr.11: 86465297-86465297 on Build GRCh38
rs761137599 Chr.11: 86465111-86465111 on Build GRCh38
rs1004656075 Chr.11: 86465403-86465403 on Build GRCh38
rs997233191 Chr.11: 86465340-86465340 on Build GRCh38
rs772738946 Chr.11: 86465086-86465086 on Build GRCh38
rs968232431 Chr.11: 86465289-86465289 on Build GRCh38
rs974021961 Chr.11: 86465074-86465074 on Build GRCh38
rs900171454 Chr.11: 86465294-86465294 on Build GRCh38
rs374593643 Chr.11: 86465113-86465113 on Build GRCh38
rs866874805 Chr.11: 86465119-86465119 on Build GRCh38
rs950553324 Chr.11: 86464989-86464989 on Build GRCh38
rs755547920 Chr.11: 86465203-86465203 on Build GRCh38
rs908621873 Chr.11: 86465027-86465027 on Build GRCh38
rs1000633643 Chr.11: 86465207-86465207 on Build GRCh38
rs774699480 Chr.11: 86465064-86465064 on Build GRCh38
rs536461690 Chr.11: 86464997-86464997 on Build GRCh38
rs199640436 Chr.11: 86465249-86465249 on Build GRCh38
rs769411375 Chr.11: 86465059-86465059 on Build GRCh38
rs765404322 Chr.11: 86465134-86465134 on Build GRCh38
rs758190862 Chr.11: 86465091-86465091 on Build GRCh38
rs950666573 Chr.11: 86465394-86465394 on Build GRCh38
rs945519534 Chr.11: 86465149-86465149 on Build GRCh38
rs954172279 Chr.11: 86465384-86465384 on Build GRCh38
rs962792951 Chr.11: 86465058-86465058 on Build GRCh38
rs1001070475 Chr.11: 86465335-86465335 on Build GRCh38
rs529230312 Chr.11: 86465391-86465391 on Build GRCh38
rs3824952 Chr.11: 86464999-86464999 on Build GRCh38
rs773663858 Chr.11: 86465095-86465095 on Build GRCh38
rs370463964 Chr.11: 86465073-86465073 on Build GRCh38
rs896256792 Chr.11: 86465238-86465238 on Build GRCh38
rs758289238 Chr.11: 86465137-86465137 on Build GRCh38
rs926605443 Chr.11: 86465426-86465426 on Build GRCh38
rs867827492 Chr.11: 86465372-86465372 on Build GRCh38
rs780909512 Chr.11: 86465050-86465050 on Build GRCh38
rs1030657210 Chr.11: 86465382-86465382 on Build GRCh38
rs372229031 Chr.11: 86465144-86465144 on Build GRCh38
rs1021088348 Chr.11: 86465266-86465266 on Build GRCh38
rs1036209758 Chr.11: 86465045-86465045 on Build GRCh38
rs745308100 Chr.11: 86465057-86465057 on Build GRCh38
rs557549197 Chr.11: 86465188-86465188 on Build GRCh38
rs780702883 Chr.11: 86465164-86465164 on Build GRCh38
rs779577786 Chr.11: 86465206-86465206 on Build GRCh38
rs554520291 Chr.11: 86465131-86465131 on Build GRCh38
rs631082 Chr.11: 86465418-86465418 on Build GRCh38
rs200546635 Chr.11: 86465126-86465126 on Build GRCh38
rs748831517 Chr.11: 86465231-86465231 on Build GRCh38
rs540188359 Chr.11: 86465273-86465273 on Build GRCh38
rs986836789 Chr.11: 86465395-86465395 on Build GRCh38
rs770356008 Chr.11: 86465447-86465447 on Build GRCh38
rs746833927 Chr.11: 86465046-86465046 on Build GRCh38
rs770520958 Chr.11: 86465047-86465047 on Build GRCh38
rs974572202 Chr.11: 86465420-86465420 on Build GRCh38
rs759524876 Chr.11: 86465127-86465127 on Build GRCh38
rs757050542 Chr.11: 86465159-86465159 on Build GRCh38

View Full Product Details