Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 110136579-110137089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H12C
Entrez Gene ID
85463
Gene Name
zinc finger CCCH-type containing 12C
Gene Aliases
MCPIP3
UniGene
Hs.376289
Gene Chromosome Location
Chr.11: 110092425-110171841 on Build GRCh38
Transcripts
NM_033390.1 XM_005271715.3 XM_011543055.2 XM_017018477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4742245 c.83G>C p.R28P Chr.11: 110136724-110136724 on Build GRCh38
5734837 c.102G>C p.L34F Chr.11: 110136743-110136743 on Build GRCh38
267965 c.239C>T p.A80V Chr.11: 110136880-110136880 on Build GRCh38
1627762 c.219G>T p.V73V Chr.11: 110136860-110136860 on Build GRCh38
6825611 c.398G>A p.R133H Chr.11: 110137039-110137039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770171952 Chr.11: 110136682-110136682 on Build GRCh38
rs752698103 Chr.11: 110136768-110136768 on Build GRCh38
rs747784239 Chr.11: 110136819-110136819 on Build GRCh38
rs552818767 Chr.11: 110136681-110136681 on Build GRCh38
rs759579168 Chr.11: 110136990-110136990 on Build GRCh38
rs780281950 Chr.11: 110136922-110136922 on Build GRCh38
rs377391499 Chr.11: 110137039-110137039 on Build GRCh38
rs761650267 Chr.11: 110136614-110136614 on Build GRCh38
rs757729109 Chr.11: 110136886-110136886 on Build GRCh38
rs371698081 Chr.11: 110136694-110136694 on Build GRCh38
rs867508273 Chr.11: 110136770-110136771 on Build GRCh38
rs201314969 Chr.11: 110136669-110136669 on Build GRCh38
rs972697950 Chr.11: 110137042-110137042 on Build GRCh38
rs748972673 Chr.11: 110137041-110137041 on Build GRCh38
rs376845158 Chr.11: 110136992-110136992 on Build GRCh38
rs775149266 Chr.11: 110136692-110136693 on Build GRCh38
rs746269536 Chr.11: 110136811-110136811 on Build GRCh38
rs760945658 Chr.11: 110137002-110137002 on Build GRCh38
rs926596847 Chr.11: 110136657-110136657 on Build GRCh38
rs972730291 Chr.11: 110136779-110136779 on Build GRCh38
rs779244027 Chr.11: 110136805-110136805 on Build GRCh38
rs965725013 Chr.11: 110136769-110136769 on Build GRCh38
rs766223495 Chr.11: 110136760-110136760 on Build GRCh38
rs776685534 Chr.11: 110136771-110136772 on Build GRCh38
rs1021129070 Chr.11: 110136683-110136683 on Build GRCh38
rs749947569 Chr.11: 110136894-110136894 on Build GRCh38
rs758916089 Chr.11: 110137024-110137024 on Build GRCh38
rs368225431 Chr.11: 110136724-110136724 on Build GRCh38
rs751462680 Chr.11: 110136761-110136761 on Build GRCh38
rs755763390 Chr.11: 110137038-110137038 on Build GRCh38
rs750161142 Chr.11: 110136656-110136656 on Build GRCh38
rs895513581 Chr.11: 110136658-110136658 on Build GRCh38
rs966935865 Chr.11: 110136699-110136699 on Build GRCh38
rs1029875059 Chr.11: 110136863-110136863 on Build GRCh38
rs760547466 Chr.11: 110136717-110136718 on Build GRCh38
rs758720104 Chr.11: 110136912-110136912 on Build GRCh38
rs780449802 Chr.11: 110137031-110137031 on Build GRCh38
rs775161683 Chr.11: 110136996-110136996 on Build GRCh38
rs764671894 Chr.11: 110136733-110136733 on Build GRCh38
rs760670937 Chr.11: 110136723-110136723 on Build GRCh38
rs985986349 Chr.11: 110136765-110136765 on Build GRCh38
rs764263261 Chr.11: 110137006-110137006 on Build GRCh38
rs760306301 Chr.11: 110136717-110136717 on Build GRCh38
rs755249395 Chr.11: 110136763-110136763 on Build GRCh38
rs190876795 Chr.11: 110136672-110136672 on Build GRCh38
rs201003221 Chr.11: 110136842-110136842 on Build GRCh38
rs764220083 Chr.11: 110136880-110136880 on Build GRCh38
rs765257532 Chr.11: 110136632-110136632 on Build GRCh38
rs1043597052 Chr.11: 110137005-110137005 on Build GRCh38
rs548799810 Chr.11: 110137019-110137019 on Build GRCh38
rs1007367602 Chr.11: 110137020-110137020 on Build GRCh38
rs757818990 Chr.11: 110136800-110136800 on Build GRCh38
rs777746568 Chr.11: 110136790-110136790 on Build GRCh38
rs773157444 Chr.11: 110136754-110136754 on Build GRCh38
rs746394139 Chr.11: 110136909-110136909 on Build GRCh38
rs200952777 Chr.11: 110136767-110136767 on Build GRCh38
rs918482173 Chr.11: 110136838-110136838 on Build GRCh38
rs138327298 Chr.11: 110136668-110136668 on Build GRCh38
rs1009818694 Chr.11: 110136906-110136906 on Build GRCh38
rs199980912 Chr.11: 110136818-110136818 on Build GRCh38
rs895575128 Chr.11: 110136898-110136898 on Build GRCh38
rs368637944 Chr.11: 110136981-110136981 on Build GRCh38
rs374918698 Chr.11: 110136852-110136852 on Build GRCh38
rs373507663 Chr.11: 110136948-110136948 on Build GRCh38
rs182300452 Chr.11: 110137018-110137018 on Build GRCh38
rs962854926 Chr.11: 110136925-110136925 on Build GRCh38
rs753929999 Chr.11: 110137009-110137009 on Build GRCh38
rs761830979 Chr.11: 110137011-110137011 on Build GRCh38
rs902745916 Chr.11: 110136927-110136927 on Build GRCh38
rs758160086 Chr.11: 110136665-110136665 on Build GRCh38
rs927144538 Chr.11: 110136858-110136858 on Build GRCh38
rs770972496 Chr.11: 110136964-110136964 on Build GRCh38
rs768638115 Chr.11: 110136752-110136753 on Build GRCh38
rs762939309 Chr.11: 110136840-110136840 on Build GRCh38
rs768598487 Chr.11: 110136934-110136934 on Build GRCh38
rs756171326 Chr.11: 110136778-110136778 on Build GRCh38
rs756296273 Chr.11: 110136865-110136865 on Build GRCh38
rs369275888 Chr.11: 110136881-110136881 on Build GRCh38
rs772607446 Chr.11: 110136837-110136837 on Build GRCh38
rs75716865 Chr.11: 110136730-110136730 on Build GRCh38
rs768273783 Chr.11: 110136814-110136814 on Build GRCh38
rs560299120 Chr.11: 110136793-110136793 on Build GRCh38
rs374188395 Chr.11: 110137032-110137032 on Build GRCh38
rs749738054 Chr.11: 110136794-110136794 on Build GRCh38
rs778011615 Chr.11: 110137023-110137023 on Build GRCh38
rs750859968 Chr.11: 110137021-110137021 on Build GRCh38
rs761922893 Chr.11: 110137018-110137019 on Build GRCh38
rs776373695 Chr.11: 110136656-110136657 on Build GRCh38
rs772778293 Chr.11: 110136941-110136941 on Build GRCh38
rs779443852 Chr.11: 110136904-110136904 on Build GRCh38
rs375346362 Chr.11: 110136759-110136759 on Build GRCh38
rs536697866 Chr.11: 110136844-110136844 on Build GRCh38
rs752925233 Chr.11: 110136857-110136857 on Build GRCh38
rs769276498 Chr.11: 110136835-110136835 on Build GRCh38
rs372870702 Chr.11: 110136851-110136851 on Build GRCh38
rs778394205 Chr.11: 110136687-110136687 on Build GRCh38
rs749782664 Chr.11: 110136689-110136689 on Build GRCh38

View Full Product Details