Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 110159174-110159672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H12C
Entrez Gene ID
85463
Gene Name
zinc finger CCCH-type containing 12C
Gene Aliases
MCPIP3
UniGene
Hs.376289
Gene Chromosome Location
Chr.11: 110092425-110171841 on Build GRCh38
Transcripts
NM_033390.1 XM_005271715.3 XM_011543055.2 XM_017018477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4742246 c.1042G>A p.G348S Chr.11: 110159384-110159384 on Build GRCh38
3808313 c.1133C>A p.S378* Chr.11: 110159475-110159475 on Build GRCh38
5361007 c.1072G>A p.D358N Chr.11: 110159414-110159414 on Build GRCh38
922899 c.1145A>G p.D382G Chr.11: 110159487-110159487 on Build GRCh38
4655178 c.1051G>A p.V351M Chr.11: 110159393-110159393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758277003 Chr.11: 110159293-110159293 on Build GRCh38
rs770133684 Chr.11: 110159379-110159379 on Build GRCh38
rs369141454 Chr.11: 110159207-110159207 on Build GRCh38
rs931554025 Chr.11: 110159397-110159397 on Build GRCh38
rs117504164 Chr.11: 110159482-110159482 on Build GRCh38
rs769349031 Chr.11: 110159239-110159239 on Build GRCh38
rs1029252013 Chr.11: 110159624-110159624 on Build GRCh38
rs921446410 Chr.11: 110159552-110159552 on Build GRCh38
rs751851502 Chr.11: 110159450-110159450 on Build GRCh38
rs745507595 Chr.11: 110159383-110159383 on Build GRCh38
rs750307348 Chr.11: 110159437-110159437 on Build GRCh38
rs1038285564 Chr.11: 110159648-110159648 on Build GRCh38
rs756641293 Chr.11: 110159517-110159517 on Build GRCh38
rs372574677 Chr.11: 110159392-110159392 on Build GRCh38
rs747122949 Chr.11: 110159348-110159348 on Build GRCh38
rs750614233 Chr.11: 110159325-110159325 on Build GRCh38
rs1030950604 Chr.11: 110159610-110159610 on Build GRCh38
rs764133995 Chr.11: 110159305-110159305 on Build GRCh38
rs557218606 Chr.11: 110159344-110159344 on Build GRCh38
rs887084959 Chr.11: 110159583-110159583 on Build GRCh38
rs373900166 Chr.11: 110159240-110159240 on Build GRCh38
rs1016879783 Chr.11: 110159226-110159226 on Build GRCh38
rs757034841 Chr.11: 110159315-110159315 on Build GRCh38
rs200240485 Chr.11: 110159271-110159271 on Build GRCh38
rs541087297 Chr.11: 110159595-110159595 on Build GRCh38
rs369911747 Chr.11: 110159259-110159259 on Build GRCh38
rs772211117 Chr.11: 110159232-110159232 on Build GRCh38
rs373315500 Chr.11: 110159270-110159270 on Build GRCh38
rs3802839 Chr.11: 110159503-110159503 on Build GRCh38
rs771692180 Chr.11: 110159542-110159542 on Build GRCh38
rs199811747 Chr.11: 110159248-110159248 on Build GRCh38
rs377608051 Chr.11: 110159353-110159353 on Build GRCh38
rs774017439 Chr.11: 110159252-110159252 on Build GRCh38
rs753739956 Chr.11: 110159309-110159309 on Build GRCh38
rs777288731 Chr.11: 110159374-110159374 on Build GRCh38
rs749695643 Chr.11: 110159521-110159521 on Build GRCh38
rs761670342 Chr.11: 110159411-110159411 on Build GRCh38
rs776563557 Chr.11: 110159402-110159402 on Build GRCh38
rs760228663 Chr.11: 110159400-110159400 on Build GRCh38
rs568952462 Chr.11: 110159380-110159380 on Build GRCh38
rs758659102 Chr.11: 110159443-110159443 on Build GRCh38
rs577220792 Chr.11: 110159582-110159582 on Build GRCh38
rs370069822 Chr.11: 110159316-110159316 on Build GRCh38
rs895276058 Chr.11: 110159488-110159488 on Build GRCh38
rs759196134 Chr.11: 110159253-110159253 on Build GRCh38
rs780293518 Chr.11: 110159235-110159235 on Build GRCh38
rs747501221 Chr.11: 110159237-110159237 on Build GRCh38
rs185400542 Chr.11: 110159211-110159211 on Build GRCh38
rs1004239216 Chr.11: 110159614-110159614 on Build GRCh38
rs906569110 Chr.11: 110159566-110159567 on Build GRCh38
rs760604390 Chr.11: 110159276-110159276 on Build GRCh38
rs780272912 Chr.11: 110159347-110159347 on Build GRCh38
rs765067151 Chr.11: 110159424-110159424 on Build GRCh38
rs778107110 Chr.11: 110159520-110159520 on Build GRCh38
rs1010786120 Chr.11: 110159617-110159617 on Build GRCh38
rs755516129 Chr.11: 110159360-110159360 on Build GRCh38
rs752637781 Chr.11: 110159250-110159250 on Build GRCh38
rs771779970 Chr.11: 110159387-110159387 on Build GRCh38
rs781138402 Chr.11: 110159484-110159484 on Build GRCh38
rs934036174 Chr.11: 110159560-110159560 on Build GRCh38
rs766754150 Chr.11: 110159449-110159449 on Build GRCh38

View Full Product Details