Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111281274-111281814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf53
Entrez Gene ID
341032
Gene Name
chromosome 11 open reading frame 53
Gene Aliases
-
UniGene
Hs.298685
Gene Chromosome Location
Chr.11: 111245720-111286406 on Build GRCh38
Transcripts
NM_198498.2 XM_011542803.2 XM_011542804.2

SNP Detail

SNP ID Chromosome Location
rs1019630281 Chr.11: 111281480-111281480 on Build GRCh38
rs377155166 Chr.11: 111281431-111281431 on Build GRCh38
rs766907032 Chr.11: 111281487-111281487 on Build GRCh38
rs1017878427 Chr.11: 111281481-111281481 on Build GRCh38
rs1026134439 Chr.11: 111281517-111281517 on Build GRCh38
rs4636709 Chr.11: 111281664-111281664 on Build GRCh38
rs755367387 Chr.11: 111281424-111281424 on Build GRCh38
rs371718344 Chr.11: 111281784-111281784 on Build GRCh38
rs751312579 Chr.11: 111281688-111281688 on Build GRCh38
rs544336438 Chr.11: 111281323-111281323 on Build GRCh38
rs889997910 Chr.11: 111281735-111281735 on Build GRCh38
rs777557568 Chr.11: 111281434-111281434 on Build GRCh38
rs184672993 Chr.11: 111281492-111281492 on Build GRCh38
rs116063175 Chr.11: 111281474-111281474 on Build GRCh38
rs908973529 Chr.11: 111281535-111281535 on Build GRCh38
rs890060859 Chr.11: 111281462-111281462 on Build GRCh38
rs904001529 Chr.11: 111281702-111281702 on Build GRCh38
rs573814977 Chr.11: 111281778-111281778 on Build GRCh38
rs775338039 Chr.11: 111281638-111281638 on Build GRCh38
rs766535281 Chr.11: 111281592-111281592 on Build GRCh38
rs772360194 Chr.11: 111281464-111281464 on Build GRCh38
rs188633452 Chr.11: 111281521-111281521 on Build GRCh38
rs12416966 Chr.11: 111281472-111281472 on Build GRCh38
rs372697986 Chr.11: 111281404-111281404 on Build GRCh38
rs913447941 Chr.11: 111281642-111281642 on Build GRCh38
rs566608445 Chr.11: 111281753-111281753 on Build GRCh38
rs200425567 Chr.11: 111281473-111281473 on Build GRCh38
rs773446874 Chr.11: 111281499-111281499 on Build GRCh38
rs878975085 Chr.11: 111281622-111281622 on Build GRCh38
rs58163628 Chr.11: 111281448-111281448 on Build GRCh38
rs979860424 Chr.11: 111281548-111281548 on Build GRCh38
rs1049935610 Chr.11: 111281732-111281732 on Build GRCh38
rs749248772 Chr.11: 111281563-111281563 on Build GRCh38
rs937937255 Chr.11: 111281656-111281656 on Build GRCh38
rs754298546 Chr.11: 111281581-111281581 on Build GRCh38
rs775354344 Chr.11: 111281459-111281459 on Build GRCh38
rs562129011 Chr.11: 111281333-111281333 on Build GRCh38
rs886327439 Chr.11: 111281740-111281740 on Build GRCh38
rs993089180 Chr.11: 111281632-111281632 on Build GRCh38
rs193152060 Chr.11: 111281690-111281690 on Build GRCh38
rs897189130 Chr.11: 111281774-111281774 on Build GRCh38
rs997263254 Chr.11: 111281719-111281719 on Build GRCh38
rs76820831 Chr.11: 111281789-111281789 on Build GRCh38
rs778513380 Chr.11: 111281441-111281441 on Build GRCh38
rs1008526119 Chr.11: 111281768-111281768 on Build GRCh38
rs994234971 Chr.11: 111281777-111281777 on Build GRCh38
rs756452162 Chr.11: 111281401-111281401 on Build GRCh38
rs370889743 Chr.11: 111281435-111281435 on Build GRCh38
rs904051823 Chr.11: 111281334-111281334 on Build GRCh38
rs774298873 Chr.11: 111281424-111281424 on Build GRCh38
rs938825935 Chr.11: 111281585-111281585 on Build GRCh38
rs945119089 Chr.11: 111281687-111281687 on Build GRCh38
rs759426670 Chr.11: 111281437-111281437 on Build GRCh38
rs773257907 Chr.11: 111281482-111281482 on Build GRCh38
rs936809858 Chr.11: 111281720-111281720 on Build GRCh38
rs938957954 Chr.11: 111281385-111281385 on Build GRCh38
rs951995426 Chr.11: 111281530-111281530 on Build GRCh38
rs1987128 Chr.11: 111281771-111281771 on Build GRCh38
rs772014487 Chr.11: 111281452-111281452 on Build GRCh38
rs758891020 Chr.11: 111281399-111281399 on Build GRCh38
rs551127578 Chr.11: 111281643-111281643 on Build GRCh38
rs191706450 Chr.11: 111281443-111281443 on Build GRCh38
rs79584066 Chr.11: 111281539-111281539 on Build GRCh38
rs780478047 Chr.11: 111281403-111281403 on Build GRCh38
rs1057392941 Chr.11: 111281400-111281400 on Build GRCh38
rs111894577 Chr.11: 111281673-111281673 on Build GRCh38
rs760504824 Chr.11: 111281466-111281466 on Build GRCh38
rs1001108634 Chr.11: 111281352-111281352 on Build GRCh38
rs762987433 Chr.11: 111281485-111281485 on Build GRCh38
rs5794734 Chr.11: 111281539-111281540 on Build GRCh38
rs368212509 Chr.11: 111281498-111281498 on Build GRCh38

View Full Product Details