Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1780359-1780859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOR2
Entrez Gene ID
79602
Gene Name
adiponectin receptor 2
Gene Aliases
ACDCR2, PAQR2
UniGene
Hs.371642
Gene Chromosome Location
Chr.12: 1690981-1788679 on Build GRCh38
Transcripts
NM_024551.2 XM_005253789.1 XM_006719018.2 XM_011521024.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4607542 c.538G>T p.E180* Chr.12: 1780525-1780525 on Build GRCh38
5777459 c.577C>G p.L193V Chr.12: 1780564-1780564 on Build GRCh38
6462465 c.626A>G p.E209G Chr.12: 1780613-1780613 on Build GRCh38
937745 c.637C>T p.R213W Chr.12: 1780624-1780624 on Build GRCh38
2003972 c.627G>C p.E209D Chr.12: 1780614-1780614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76885456 Chr.12: 1780589-1780589 on Build GRCh38
rs562288254 Chr.12: 1780678-1780678 on Build GRCh38
rs201727993 Chr.12: 1780564-1780564 on Build GRCh38
rs748585007 Chr.12: 1780631-1780631 on Build GRCh38
rs915604361 Chr.12: 1780801-1780801 on Build GRCh38
rs756843147 Chr.12: 1780571-1780571 on Build GRCh38
rs114675341 Chr.12: 1780766-1780766 on Build GRCh38
rs79920875 Chr.12: 1780789-1780789 on Build GRCh38
rs551852718 Chr.12: 1780652-1780652 on Build GRCh38
rs764160007 Chr.12: 1780545-1780545 on Build GRCh38
rs764519900 Chr.12: 1780743-1780743 on Build GRCh38
rs773243816 Chr.12: 1780466-1780466 on Build GRCh38
rs10848575 Chr.12: 1780754-1780754 on Build GRCh38
rs907079041 Chr.12: 1780400-1780400 on Build GRCh38
rs757956906 Chr.12: 1780686-1780686 on Build GRCh38
rs1013161634 Chr.12: 1780385-1780385 on Build GRCh38
rs766535675 Chr.12: 1780476-1780476 on Build GRCh38
rs748587883 Chr.12: 1780435-1780435 on Build GRCh38
rs749305937 Chr.12: 1780647-1780647 on Build GRCh38
rs182238128 Chr.12: 1780794-1780794 on Build GRCh38
rs767870 Chr.12: 1780657-1780657 on Build GRCh38
rs1045942705 Chr.12: 1780806-1780806 on Build GRCh38
rs770243021 Chr.12: 1780638-1780638 on Build GRCh38
rs764066042 Chr.12: 1780666-1780666 on Build GRCh38
rs749997156 Chr.12: 1780684-1780684 on Build GRCh38
rs553727192 Chr.12: 1780617-1780617 on Build GRCh38
rs998783620 Chr.12: 1780407-1780407 on Build GRCh38
rs773726684 Chr.12: 1780644-1780644 on Build GRCh38
rs770999657 Chr.12: 1780648-1780648 on Build GRCh38
rs755978842 Chr.12: 1780531-1780531 on Build GRCh38
rs767205765 Chr.12: 1780493-1780493 on Build GRCh38
rs200439501 Chr.12: 1780753-1780753 on Build GRCh38
rs972270406 Chr.12: 1780703-1780703 on Build GRCh38
rs948504422 Chr.12: 1780805-1780805 on Build GRCh38
rs778643792 Chr.12: 1780593-1780593 on Build GRCh38
rs760569084 Chr.12: 1780662-1780662 on Build GRCh38
rs775110533 Chr.12: 1780658-1780658 on Build GRCh38
rs753523903 Chr.12: 1780591-1780592 on Build GRCh38
rs74059364 Chr.12: 1780687-1780687 on Build GRCh38
rs756136615 Chr.12: 1780401-1780401 on Build GRCh38
rs374098501 Chr.12: 1780404-1780404 on Build GRCh38
rs985679669 Chr.12: 1780554-1780554 on Build GRCh38
rs768908704 Chr.12: 1780419-1780419 on Build GRCh38
rs377630854 Chr.12: 1780424-1780424 on Build GRCh38
rs953529863 Chr.12: 1780671-1780671 on Build GRCh38
rs770317650 Chr.12: 1780800-1780800 on Build GRCh38
rs537605834 Chr.12: 1780776-1780776 on Build GRCh38
rs186366553 Chr.12: 1780683-1780683 on Build GRCh38
rs999992104 Chr.12: 1780767-1780767 on Build GRCh38
rs372747022 Chr.12: 1780744-1780744 on Build GRCh38
rs774447126 Chr.12: 1780480-1780480 on Build GRCh38
rs770334866 Chr.12: 1780447-1780447 on Build GRCh38
rs548839495 Chr.12: 1780756-1780756 on Build GRCh38
rs373789695 Chr.12: 1780669-1780669 on Build GRCh38
rs1030981776 Chr.12: 1780581-1780581 on Build GRCh38
rs747342627 Chr.12: 1780418-1780418 on Build GRCh38
rs759873192 Chr.12: 1780485-1780485 on Build GRCh38
rs377651133 Chr.12: 1780387-1780387 on Build GRCh38
rs879717049 Chr.12: 1780754-1780755 on Build GRCh38
rs966238035 Chr.12: 1780697-1780697 on Build GRCh38
rs199835605 Chr.12: 1780428-1780428 on Build GRCh38
rs763047459 Chr.12: 1780475-1780475 on Build GRCh38
rs777739515 Chr.12: 1780417-1780418 on Build GRCh38
rs745501992 Chr.12: 1780601-1780601 on Build GRCh38
rs71450619 Chr.12: 1780754-1780755 on Build GRCh38
rs575657654 Chr.12: 1780754-1780755 on Build GRCh38
rs753350785 Chr.12: 1780566-1780566 on Build GRCh38
rs562879128 Chr.12: 1780726-1780726 on Build GRCh38
rs752590307 Chr.12: 1780515-1780515 on Build GRCh38
rs2887630 Chr.12: 1780755-1780755 on Build GRCh38
rs893922616 Chr.12: 1780784-1780784 on Build GRCh38
rs759657714 Chr.12: 1780656-1780656 on Build GRCh38
rs1024144024 Chr.12: 1780392-1780392 on Build GRCh38
rs757052625 Chr.12: 1780672-1780673 on Build GRCh38
rs533461747 Chr.12: 1780738-1780738 on Build GRCh38
rs770667901 Chr.12: 1780413-1780413 on Build GRCh38
rs375777748 Chr.12: 1780822-1780822 on Build GRCh38
rs895734056 Chr.12: 1780744-1780744 on Build GRCh38

View Full Product Details