Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 3648227-3648740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRACR2A
Entrez Gene ID
84766
Gene Name
calcium release activated channel regulator 2A
Gene Aliases
EFCAB4B
UniGene
Hs.504534
Gene Chromosome Location
Chr.12: 3613366-3753216 on Build GRCh38
Transcripts
NM_001144958.1 NM_032680.3 XM_006719021.3 XM_011521034.2 XM_011521036.2 XM_011521037.2 XM_011521038.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3811908 c.1109A>G p.D370G Chr.12: 3648551-3648551 on Build GRCh38
6519879 c.1086G>T p.K362N Chr.12: 3648574-3648574 on Build GRCh38
2206412 c.1118+4C>T p.? Chr.12: 3648538-3648538 on Build GRCh38
6420247 c.1097T>C p.L366P Chr.12: 3648563-3648563 on Build GRCh38
3460350 c.1113C>T p.F371F Chr.12: 3648547-3648547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201378260 Chr.12: 3648570-3648570 on Build GRCh38
rs769781947 Chr.12: 3648538-3648538 on Build GRCh38
rs140018162 Chr.12: 3648488-3648488 on Build GRCh38
rs1049931601 Chr.12: 3648383-3648383 on Build GRCh38
rs71534261 Chr.12: 3648355-3648355 on Build GRCh38
rs536808904 Chr.12: 3648428-3648428 on Build GRCh38
rs372821272 Chr.12: 3648596-3648596 on Build GRCh38
rs946850940 Chr.12: 3648425-3648425 on Build GRCh38
rs745965433 Chr.12: 3648442-3648442 on Build GRCh38
rs981053822 Chr.12: 3648286-3648286 on Build GRCh38
rs74055951 Chr.12: 3648455-3648455 on Build GRCh38
rs780599403 Chr.12: 3648510-3648510 on Build GRCh38
rs977274463 Chr.12: 3648357-3648357 on Build GRCh38
rs770534591 Chr.12: 3648578-3648578 on Build GRCh38
rs877554 Chr.12: 3648258-3648258 on Build GRCh38
rs755918542 Chr.12: 3648624-3648624 on Build GRCh38
rs750735904 Chr.12: 3648441-3648441 on Build GRCh38
rs926587881 Chr.12: 3648305-3648305 on Build GRCh38
rs561395966 Chr.12: 3648595-3648595 on Build GRCh38
rs748740997 Chr.12: 3648577-3648577 on Build GRCh38
rs888140992 Chr.12: 3648676-3648676 on Build GRCh38
rs143542484 Chr.12: 3648535-3648535 on Build GRCh38
rs749083509 Chr.12: 3648598-3648599 on Build GRCh38
rs952762130 Chr.12: 3648344-3648344 on Build GRCh38
rs961543427 Chr.12: 3648568-3648568 on Build GRCh38
rs201245194 Chr.12: 3648467-3648467 on Build GRCh38
rs567342642 Chr.12: 3648720-3648720 on Build GRCh38
rs370367338 Chr.12: 3648537-3648537 on Build GRCh38
rs34594218 Chr.12: 3648447-3648447 on Build GRCh38
rs922602049 Chr.12: 3648368-3648368 on Build GRCh38
rs908598635 Chr.12: 3648348-3648348 on Build GRCh38
rs79670551 Chr.12: 3648454-3648454 on Build GRCh38
rs531444591 Chr.12: 3648667-3648667 on Build GRCh38
rs778537371 Chr.12: 3648638-3648638 on Build GRCh38
rs777676733 Chr.12: 3648627-3648627 on Build GRCh38
rs764455333 Chr.12: 3648436-3648436 on Build GRCh38
rs76430563 Chr.12: 3648650-3648650 on Build GRCh38
rs780324912 Chr.12: 3648453-3648453 on Build GRCh38
rs948347124 Chr.12: 3648670-3648670 on Build GRCh38
rs1050379995 Chr.12: 3648442-3648443 on Build GRCh38
rs777446938 Chr.12: 3648514-3648528 on Build GRCh38
rs149287496 Chr.12: 3648487-3648487 on Build GRCh38
rs910460391 Chr.12: 3648625-3648625 on Build GRCh38
rs527267393 Chr.12: 3648701-3648701 on Build GRCh38
rs759183346 Chr.12: 3648253-3648253 on Build GRCh38
rs755180141 Chr.12: 3648608-3648608 on Build GRCh38
rs757786786 Chr.12: 3648492-3648492 on Build GRCh38
rs749293414 Chr.12: 3648632-3648632 on Build GRCh38
rs750458814 Chr.12: 3648520-3648520 on Build GRCh38
rs551652517 Chr.12: 3648668-3648668 on Build GRCh38
rs752338437 Chr.12: 3648301-3648301 on Build GRCh38
rs376963180 Chr.12: 3648622-3648622 on Build GRCh38
rs148020723 Chr.12: 3648571-3648571 on Build GRCh38
rs141677632 Chr.12: 3648611-3648611 on Build GRCh38
rs1053416953 Chr.12: 3648468-3648468 on Build GRCh38
rs940122458 Chr.12: 3648380-3648380 on Build GRCh38
rs1009272725 Chr.12: 3648507-3648507 on Build GRCh38
rs745398108 Chr.12: 3648640-3648640 on Build GRCh38
rs887304 Chr.12: 3648382-3648382 on Build GRCh38
rs773874892 Chr.12: 3648581-3648581 on Build GRCh38
rs765872014 Chr.12: 3648514-3648514 on Build GRCh38
rs1005907767 Chr.12: 3648546-3648546 on Build GRCh38
rs142666646 Chr.12: 3648318-3648318 on Build GRCh38
rs140483672 Chr.12: 3648660-3648660 on Build GRCh38
rs12308840 Chr.12: 3648702-3648702 on Build GRCh38
rs751023994 Chr.12: 3648601-3648601 on Build GRCh38
rs150013368 Chr.12: 3648673-3648673 on Build GRCh38
rs573814749 Chr.12: 3648385-3648385 on Build GRCh38
rs908616005 Chr.12: 3648356-3648356 on Build GRCh38
rs758649126 Chr.12: 3648445-3648445 on Build GRCh38
rs146788034 Chr.12: 3648659-3648659 on Build GRCh38
rs762481615 Chr.12: 3648483-3648483 on Build GRCh38
rs765778555 Chr.12: 3648600-3648600 on Build GRCh38
rs142731912 Chr.12: 3648378-3648379 on Build GRCh38
rs751708113 Chr.12: 3648603-3648603 on Build GRCh38
rs374054088 Chr.12: 3648556-3648556 on Build GRCh38
rs970932917 Chr.12: 3648529-3648529 on Build GRCh38
rs764589720 Chr.12: 3648491-3648491 on Build GRCh38
rs371628713 Chr.12: 3648480-3648480 on Build GRCh38
rs374515541 Chr.12: 3648565-3648565 on Build GRCh38
rs770348652 Chr.12: 3648664-3648664 on Build GRCh38
rs758562218 Chr.12: 3648534-3648534 on Build GRCh38
rs545817495 Chr.12: 3648582-3648582 on Build GRCh38
rs773139580 Chr.12: 3648572-3648572 on Build GRCh38
rs368326307 Chr.12: 3648637-3648637 on Build GRCh38
rs754429567 Chr.12: 3648439-3648439 on Build GRCh38
rs368297120 Chr.12: 3648504-3648504 on Build GRCh38
rs915506692 Chr.12: 3648395-3648395 on Build GRCh38
rs1009681532 Chr.12: 3648252-3648252 on Build GRCh38

View Full Product Details