Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6492067-6492573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL51
Entrez Gene ID
51258
Gene Name
mitochondrial ribosomal protein L51
Gene Aliases
CDA09, HSPC241, MRP64, bMRP64
UniGene
Hs.55847
Gene Chromosome Location
Chr.12: 6492150-6493305 on Build GRCh38
Transcripts
NM_016497.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1991610 c.354C>A p.Y118* Chr.12: 6492304-6492304 on Build GRCh38
6715420 c.268C>T p.R90C Chr.12: 6492390-6492390 on Build GRCh38
179482 c.382C>T p.R128* Chr.12: 6492276-6492276 on Build GRCh38
6715419 c.310G>A p.A104T Chr.12: 6492348-6492348 on Build GRCh38
7111888 c.309C>A p.F103L Chr.12: 6492349-6492349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61570300 Chr.12: 6492525-6492525 on Build GRCh38
rs202063773 Chr.12: 6492258-6492258 on Build GRCh38
rs1002324800 Chr.12: 6492466-6492466 on Build GRCh38
rs756465282 Chr.12: 6492507-6492507 on Build GRCh38
rs1006105633 Chr.12: 6492188-6492188 on Build GRCh38
rs371441476 Chr.12: 6492442-6492442 on Build GRCh38
rs771957365 Chr.12: 6492519-6492520 on Build GRCh38
rs749456267 Chr.12: 6492512-6492512 on Build GRCh38
rs555793395 Chr.12: 6492191-6492191 on Build GRCh38
rs954320496 Chr.12: 6492318-6492318 on Build GRCh38
rs112821786 Chr.12: 6492165-6492165 on Build GRCh38
rs141891222 Chr.12: 6492453-6492453 on Build GRCh38
rs777203393 Chr.12: 6492277-6492277 on Build GRCh38
rs776718738 Chr.12: 6492275-6492275 on Build GRCh38
rs757304160 Chr.12: 6492503-6492504 on Build GRCh38
rs770882368 Chr.12: 6492519-6492519 on Build GRCh38
rs140610566 Chr.12: 6492463-6492463 on Build GRCh38
rs987141202 Chr.12: 6492330-6492330 on Build GRCh38
rs781280216 Chr.12: 6492359-6492359 on Build GRCh38
rs923102481 Chr.12: 6492185-6492185 on Build GRCh38
rs758764876 Chr.12: 6492460-6492460 on Build GRCh38
rs746422940 Chr.12: 6492233-6492233 on Build GRCh38
rs541740810 Chr.12: 6492315-6492315 on Build GRCh38
rs768645415 Chr.12: 6492356-6492356 on Build GRCh38
rs1017125758 Chr.12: 6492192-6492192 on Build GRCh38
rs774092384 Chr.12: 6492456-6492463 on Build GRCh38
rs750781979 Chr.12: 6492331-6492331 on Build GRCh38
rs766958465 Chr.12: 6492429-6492429 on Build GRCh38
rs943115261 Chr.12: 6492417-6492417 on Build GRCh38
rs770029749 Chr.12: 6492370-6492370 on Build GRCh38
rs748824105 Chr.12: 6492427-6492427 on Build GRCh38
rs373802519 Chr.12: 6492365-6492365 on Build GRCh38
rs780550355 Chr.12: 6492349-6492349 on Build GRCh38
rs370382524 Chr.12: 6492390-6492390 on Build GRCh38
rs774436320 Chr.12: 6492404-6492404 on Build GRCh38
rs755484803 Chr.12: 6492479-6492479 on Build GRCh38
rs763721706 Chr.12: 6492451-6492451 on Build GRCh38
rs747076308 Chr.12: 6492257-6492257 on Build GRCh38
rs773123494 Chr.12: 6492283-6492283 on Build GRCh38
rs758770414 Chr.12: 6492276-6492276 on Build GRCh38
rs191438555 Chr.12: 6492320-6492320 on Build GRCh38
rs1014292193 Chr.12: 6492096-6492096 on Build GRCh38
rs544027647 Chr.12: 6492110-6492110 on Build GRCh38
rs779296628 Chr.12: 6492221-6492221 on Build GRCh38
rs773211461 Chr.12: 6492382-6492382 on Build GRCh38
rs377366260 Chr.12: 6492290-6492290 on Build GRCh38
rs781173608 Chr.12: 6492487-6492487 on Build GRCh38
rs766417875 Chr.12: 6492292-6492292 on Build GRCh38
rs566101541 Chr.12: 6492538-6492538 on Build GRCh38
rs1053083822 Chr.12: 6492540-6492540 on Build GRCh38
rs753429240 Chr.12: 6492452-6492452 on Build GRCh38
rs545444196 Chr.12: 6492203-6492203 on Build GRCh38
rs1015284943 Chr.12: 6492125-6492129 on Build GRCh38
rs3948654 Chr.12: 6492121-6492121 on Build GRCh38
rs895740787 Chr.12: 6492549-6492549 on Build GRCh38
rs780173339 Chr.12: 6492250-6492250 on Build GRCh38
rs770496987 Chr.12: 6492454-6492454 on Build GRCh38
rs141081788 Chr.12: 6492348-6492348 on Build GRCh38
rs758984520 Chr.12: 6492298-6492298 on Build GRCh38
rs373380080 Chr.12: 6492476-6492476 on Build GRCh38
rs934567338 Chr.12: 6492533-6492533 on Build GRCh38
rs751174823 Chr.12: 6492297-6492297 on Build GRCh38
rs377340253 Chr.12: 6492389-6492389 on Build GRCh38
rs11380 Chr.12: 6492309-6492309 on Build GRCh38
rs925349073 Chr.12: 6492289-6492289 on Build GRCh38
rs766687025 Chr.12: 6492474-6492474 on Build GRCh38
rs768720556 Chr.12: 6492274-6492274 on Build GRCh38
rs752213313 Chr.12: 6492431-6492431 on Build GRCh38
rs558994607 Chr.12: 6492321-6492321 on Build GRCh38
rs760310024 Chr.12: 6492447-6492447 on Build GRCh38
rs558930437 Chr.12: 6492235-6492235 on Build GRCh38
rs144091628 Chr.12: 6492093-6492093 on Build GRCh38
rs9526 Chr.12: 6492352-6492352 on Build GRCh38
rs148689307 Chr.12: 6492153-6492153 on Build GRCh38
rs961969381 Chr.12: 6492133-6492133 on Build GRCh38
rs748259699 Chr.12: 6492493-6492493 on Build GRCh38
rs145116059 Chr.12: 6492353-6492353 on Build GRCh38
rs759477625 Chr.12: 6492416-6492416 on Build GRCh38

View Full Product Details