Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6731612-6732051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS7A
Entrez Gene ID
50813
Gene Name
COP9 signalosome subunit 7A
Gene Aliases
CSN7, CSN7A, SGN7a
UniGene
Hs.530823
Gene Chromosome Location
Chr.12: 6723984-6731875 on Build GRCh38
Transcripts
NM_001164093.1 NM_001164094.1 NM_001164095.2 NM_016319.3 XM_005253694.2

SNP Detail

SNP ID Chromosome Location
rs374090676 Chr.12: 6732013-6732013 on Build GRCh38
rs1008030814 Chr.12: 6731803-6731803 on Build GRCh38
rs970901851 Chr.12: 6731638-6731638 on Build GRCh38
rs181494955 Chr.12: 6731831-6731831 on Build GRCh38
rs1033813623 Chr.12: 6731926-6731926 on Build GRCh38
rs112468500 Chr.12: 6731787-6731788 on Build GRCh38
rs921222954 Chr.12: 6731832-6731832 on Build GRCh38
rs958061241 Chr.12: 6731864-6731864 on Build GRCh38
rs982716999 Chr.12: 6731646-6731646 on Build GRCh38
rs117018880 Chr.12: 6731658-6731658 on Build GRCh38
rs982222830 Chr.12: 6731908-6731908 on Build GRCh38
rs1014439402 Chr.12: 6731765-6731765 on Build GRCh38
rs983001192 Chr.12: 6731810-6731810 on Build GRCh38
rs773064912 Chr.12: 6731817-6731820 on Build GRCh38
rs965617842 Chr.12: 6731815-6731815 on Build GRCh38
rs1046974916 Chr.12: 6731771-6731771 on Build GRCh38
rs774867259 Chr.12: 6731657-6731657 on Build GRCh38
rs932265603 Chr.12: 6731843-6731843 on Build GRCh38
rs572696657 Chr.12: 6731857-6731857 on Build GRCh38
rs777146930 Chr.12: 6731901-6731901 on Build GRCh38
rs868667062 Chr.12: 6731752-6731752 on Build GRCh38
rs1010005698 Chr.12: 6731872-6731872 on Build GRCh38
rs970021864 Chr.12: 6731785-6731786 on Build GRCh38
rs761339307 Chr.12: 6731972-6731972 on Build GRCh38
rs1032460920 Chr.12: 6731846-6731846 on Build GRCh38
rs574519496 Chr.12: 6731996-6731996 on Build GRCh38
rs934442501 Chr.12: 6731863-6731863 on Build GRCh38
rs895970896 Chr.12: 6731880-6731881 on Build GRCh38
rs1040915451 Chr.12: 6731812-6731812 on Build GRCh38
rs950845317 Chr.12: 6731801-6731801 on Build GRCh38
rs138174526 Chr.12: 6731898-6731898 on Build GRCh38
rs370111243 Chr.12: 6731675-6731675 on Build GRCh38
rs994769230 Chr.12: 6731986-6731986 on Build GRCh38
rs958416096 Chr.12: 6731928-6731928 on Build GRCh38
rs78291718 Chr.12: 6732009-6732009 on Build GRCh38
rs919602264 Chr.12: 6731740-6731740 on Build GRCh38
rs999394550 Chr.12: 6731668-6731668 on Build GRCh38
rs1021194033 Chr.12: 6731887-6731887 on Build GRCh38
rs763677783 Chr.12: 6731786-6731786 on Build GRCh38
rs147181956 Chr.12: 6732000-6732000 on Build GRCh38
rs991193099 Chr.12: 6731968-6731968 on Build GRCh38
rs760175996 Chr.12: 6731763-6731763 on Build GRCh38
rs113935173 Chr.12: 6731780-6731780 on Build GRCh38
rs919712560 Chr.12: 6731982-6731982 on Build GRCh38
rs896031041 Chr.12: 6731706-6731706 on Build GRCh38
rs996940763 Chr.12: 6731844-6731844 on Build GRCh38
rs1027662631 Chr.12: 6731797-6731797 on Build GRCh38
rs143732908 Chr.12: 6731984-6731984 on Build GRCh38

View Full Product Details