Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6825385-6825886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR162
Entrez Gene ID
27239
Gene Name
G protein-coupled receptor 162
Gene Aliases
A-2, GRCA
UniGene
Hs.631654
Gene Chromosome Location
Chr.12: 6821797-6827418 on Build GRCh38
Transcripts
NM_014449.1 NM_019858.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660098 c.895G>C p.D299H Chr.12: 6825511-6825511 on Build GRCh38
6552992 c.994G>T p.A332S Chr.12: 6825610-6825610 on Build GRCh38
4688087 c.950C>T p.T317M Chr.12: 6825566-6825566 on Build GRCh38
1606551 c.1057+6A>T p.? Chr.12: 6825679-6825679 on Build GRCh38
117583 c.935G>A p.C312Y Chr.12: 6825551-6825551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782450393 Chr.12: 6825416-6825416 on Build GRCh38
rs752085736 Chr.12: 6825489-6825489 on Build GRCh38
rs782447176 Chr.12: 6825540-6825540 on Build GRCh38
rs782715686 Chr.12: 6825597-6825597 on Build GRCh38
rs782051777 Chr.12: 6825566-6825567 on Build GRCh38
rs782197731 Chr.12: 6825606-6825606 on Build GRCh38
rs782131782 Chr.12: 6825452-6825452 on Build GRCh38
rs778444639 Chr.12: 6825769-6825769 on Build GRCh38
rs782274118 Chr.12: 6825754-6825754 on Build GRCh38
rs71445129 Chr.12: 6825479-6825479 on Build GRCh38
rs943577304 Chr.12: 6825567-6825567 on Build GRCh38
rs987707355 Chr.12: 6825767-6825767 on Build GRCh38
rs557263215 Chr.12: 6825612-6825612 on Build GRCh38
rs782379612 Chr.12: 6825487-6825487 on Build GRCh38
rs782599679 Chr.12: 6825713-6825713 on Build GRCh38
rs754497366 Chr.12: 6825736-6825738 on Build GRCh38
rs782798823 Chr.12: 6825672-6825672 on Build GRCh38
rs781904876 Chr.12: 6825535-6825535 on Build GRCh38
rs1043190991 Chr.12: 6825833-6825833 on Build GRCh38
rs781871861 Chr.12: 6825460-6825460 on Build GRCh38
rs146026728 Chr.12: 6825619-6825619 on Build GRCh38
rs782655063 Chr.12: 6825571-6825571 on Build GRCh38
rs782269584 Chr.12: 6825573-6825573 on Build GRCh38
rs782538327 Chr.12: 6825671-6825671 on Build GRCh38
rs115573202 Chr.12: 6825464-6825464 on Build GRCh38
rs768037896 Chr.12: 6825510-6825510 on Build GRCh38
rs56390107 Chr.12: 6825415-6825415 on Build GRCh38
rs782254724 Chr.12: 6825798-6825798 on Build GRCh38
rs782010757 Chr.12: 6825492-6825492 on Build GRCh38
rs781827118 Chr.12: 6825560-6825560 on Build GRCh38
rs781934229 Chr.12: 6825507-6825507 on Build GRCh38
rs148277722 Chr.12: 6825639-6825639 on Build GRCh38
rs781839944 Chr.12: 6825528-6825528 on Build GRCh38
rs150340680 Chr.12: 6825515-6825515 on Build GRCh38
rs782249428 Chr.12: 6825615-6825615 on Build GRCh38
rs372333687 Chr.12: 6825467-6825467 on Build GRCh38
rs200768010 Chr.12: 6825501-6825501 on Build GRCh38
rs782230530 Chr.12: 6825717-6825717 on Build GRCh38
rs972174919 Chr.12: 6825735-6825735 on Build GRCh38
rs782647041 Chr.12: 6825458-6825458 on Build GRCh38
rs936417699 Chr.12: 6825850-6825850 on Build GRCh38
rs200776682 Chr.12: 6825648-6825648 on Build GRCh38
rs369299686 Chr.12: 6825524-6825524 on Build GRCh38
rs2071080 Chr.12: 6825824-6825824 on Build GRCh38
rs199552793 Chr.12: 6825585-6825585 on Build GRCh38
rs782236900 Chr.12: 6825480-6825480 on Build GRCh38
rs782133434 Chr.12: 6825519-6825519 on Build GRCh38
rs763317910 Chr.12: 6825472-6825473 on Build GRCh38
rs782162794 Chr.12: 6825620-6825620 on Build GRCh38
rs782484879 Chr.12: 6825561-6825561 on Build GRCh38
rs113976597 Chr.12: 6825462-6825462 on Build GRCh38
rs782347698 Chr.12: 6825472-6825472 on Build GRCh38
rs782502176 Chr.12: 6825723-6825723 on Build GRCh38
rs61461797 Chr.12: 6825830-6825830 on Build GRCh38
rs781818924 Chr.12: 6825435-6825435 on Build GRCh38
rs782482464 Chr.12: 6825451-6825451 on Build GRCh38
rs910775427 Chr.12: 6825804-6825804 on Build GRCh38
rs782688659 Chr.12: 6825465-6825465 on Build GRCh38
rs537356928 Chr.12: 6825494-6825494 on Build GRCh38
rs782695526 Chr.12: 6825424-6825424 on Build GRCh38
rs782444497 Chr.12: 6825690-6825690 on Build GRCh38
rs187530132 Chr.12: 6825848-6825848 on Build GRCh38
rs782385606 Chr.12: 6825616-6825616 on Build GRCh38
rs981246778 Chr.12: 6825453-6825453 on Build GRCh38
rs919254228 Chr.12: 6825739-6825739 on Build GRCh38
rs374564996 Chr.12: 6825685-6825685 on Build GRCh38
rs782721569 Chr.12: 6825556-6825556 on Build GRCh38
rs782034322 Chr.12: 6825753-6825753 on Build GRCh38
rs963458003 Chr.12: 6825707-6825707 on Build GRCh38
rs376678235 Chr.12: 6825613-6825613 on Build GRCh38
rs149471201 Chr.12: 6825633-6825633 on Build GRCh38
rs782191370 Chr.12: 6825466-6825466 on Build GRCh38

View Full Product Details