Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10418793-10419293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLRC3
Entrez Gene ID
3823
Gene Name
killer cell lectin like receptor C3
Gene Aliases
NKG2-E, NKG2E
UniGene
Hs.654362
Gene Chromosome Location
Chr.12: 10412315-10420595 on Build GRCh38
Transcripts
NM_002261.2 NM_007333.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6287330 c.287-7A>G p.? Chr.12: 10419095-10419095 on Build GRCh38
4146492 c.324C>G p.T108T Chr.12: 10419051-10419051 on Build GRCh38
4146493 c.316G>A p.A106T Chr.12: 10419059-10419059 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs11053795 Chr.12: 10419146-10419146 on Build GRCh38
rs776056734 Chr.12: 10419049-10419049 on Build GRCh38
rs968238648 Chr.12: 10418932-10418932 on Build GRCh38
rs190023003 Chr.12: 10419110-10419110 on Build GRCh38
rs28626640 Chr.12: 10419059-10419059 on Build GRCh38
rs60777328 Chr.12: 10419209-10419209 on Build GRCh38
rs4579992 Chr.12: 10418905-10418905 on Build GRCh38
rs182561471 Chr.12: 10418985-10418985 on Build GRCh38
rs36058968 Chr.12: 10418827-10418827 on Build GRCh38
rs892251981 Chr.12: 10418866-10418866 on Build GRCh38
rs1018685958 Chr.12: 10418840-10418840 on Build GRCh38
rs981060149 Chr.12: 10419229-10419229 on Build GRCh38
rs778632496 Chr.12: 10419064-10419064 on Build GRCh38
rs34487318 Chr.12: 10418880-10418880 on Build GRCh38
rs771044436 Chr.12: 10419119-10419119 on Build GRCh38
rs2682511 Chr.12: 10419117-10419117 on Build GRCh38
rs552308806 Chr.12: 10419095-10419095 on Build GRCh38
rs548611095 Chr.12: 10419130-10419130 on Build GRCh38
rs568342473 Chr.12: 10419195-10419195 on Build GRCh38
rs5796393 Chr.12: 10418937-10418938 on Build GRCh38
rs543880112 Chr.12: 10419037-10419037 on Build GRCh38
rs4275705 Chr.12: 10418915-10418915 on Build GRCh38
rs762958002 Chr.12: 10418854-10418854 on Build GRCh38
rs759868835 Chr.12: 10419047-10419047 on Build GRCh38
rs771103900 Chr.12: 10419043-10419043 on Build GRCh38
rs33947579 Chr.12: 10418940-10418941 on Build GRCh38
rs753586188 Chr.12: 10419054-10419054 on Build GRCh38
rs193205490 Chr.12: 10419227-10419227 on Build GRCh38
rs60351539 Chr.12: 10419231-10419231 on Build GRCh38
rs765628583 Chr.12: 10419048-10419048 on Build GRCh38
rs772179769 Chr.12: 10419042-10419042 on Build GRCh38
rs760290984 Chr.12: 10419052-10419052 on Build GRCh38
rs4093355 Chr.12: 10419023-10419023 on Build GRCh38
rs758206538 Chr.12: 10419098-10419098 on Build GRCh38
rs28521176 Chr.12: 10419051-10419051 on Build GRCh38
rs751020980 Chr.12: 10418831-10418831 on Build GRCh38
rs201384900 Chr.12: 10419241-10419242 on Build GRCh38
rs564052938 Chr.12: 10418969-10418969 on Build GRCh38
rs34617941 Chr.12: 10418824-10418824 on Build GRCh38
rs776863781 Chr.12: 10419045-10419045 on Build GRCh38

View Full Product Details